Merck
  • Combined derivatization and high-performance liquid chromatography with fluorescence and ultraviolet detection for simultaneous analysis of octreotide and gabexate mesylate metabolite in human pancreatic juice samples.
Combined derivatization and high-performance liquid chromatography with fluorescence and ultraviolet detection for simultaneous analysis of octreotide and gabexate mesylate metabolite in human pancreatic juice samples.
Biomedical chromatography : BMC (2014-10-31)
Giuseppe Carlucci, Federico Selvaggi, Sara Sulpizio, Claudio Bassi, Maura Carlucci, Roberto Cotellese, Vincenzo Ferrone, Paolo Innocenti, Marcello Locatelli
ZUSAMMENFASSUNG
A simple and sensitive method based on the combination of derivatization and high-performance liquid chromatography with ultraviolet and fluorimetric detection was developed for the simultaneous determination of octreotide and gabexate mesylate metabolite in human pancreatic juice samples. Parameters of the derivatization procedure affecting extraction efficiency were optimized. The developed method was validated according to the International Conference on Harmonization guidelines. The calibration curves were linear over a range of 0.1-15 µg/mL for octreotide and 0.20-15 µg/mL for gabexate mesylate metabolite. Derivatized products of octreotide and gabexate mesylate metabolite were separated on a Luna C18 column (4.6 × 250 mm; 5 µm particle size) using a gradient with a run time of 36 min, without further purification. The limits of detection were 0.025 and 0.05, respectively, for octreotide and gabexate mesylate metabolite. This paper reports the validation of a quantitative high performance liquid chromatography-photodiode array-fluorescence (HPLC-PDA-FL) method for the simultaneous analysis of octreotide and gabexate mesylate metabolite in pancreatic juice by protein precipitation using zinc sulfate-methanol-acetonitrile containing the derivatizing reagent, 4-fluoro-7-nitro-[2,1,3]-benzoxadiazole (NBD-F). Derivatized products of octreotide and gabexate mesylate metabolite were separated on a Luna C18 column (4.6 × 250 mm; 5 µm particle size) using a gradient with a run time of 36 min, without further purification. The method was validated over the concentration ranges 0.1-15 and 0.2-15 µg/mL for octreotide and gabexate mesylate metabolite, respectively, in human pancreatic juice. Biphalin and methyl-p-hydroxybenzoate were used as the internal standards. This method was successfully utilized to support clinical studies in humans. The results from assay validations show that the method is selective, sensitive and robust. The limit of quantification of the method was 0.1 µg/mL for octreotide and 0.2 µg/mL for gabexate mesylate metabolite, and matrix matched standard curves showed a good linearity up to 15 µg/mL. In the entire analytical range the intra- and inter-day precision (RSD%) values were respectively ≤5.9% and ≤3.1% for octreotide and ≤2.0% and ≤3.9% for gabexate mesylate metabolite. For both analytes the intra- and inter-day accuracy (bias) values ranged respectively from -6.8 to -2.5% and from -4.6 to -5.7%. This method utilizes derivatization with NBD-F and provides adequate sensitivity for both drugs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Phthaldialdehyd, ≥97% (HPLC), powder or crystals
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Methyl-4-hydroxybenzoat, ReagentPlus®, ≥99.0%, crystalline
Sigma-Aldrich
Phthaldialdehyd, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Phthaldialdehyd, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder or crystals
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
Methyl-4-hydroxybenzoat, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Octylphosphonic acid, 97%
Sigma-Aldrich
Methyl-4-hydroxybenzoat, ≥99%, FCC
Sigma-Aldrich
Octreotide, ≥98% (HPLC)
Sigma-Aldrich
Methyl-4-hydroxybenzoat, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methyl-4-hydroxybenzoat, analytical standard
Sigma-Aldrich
4-Fluor-7-nitrobenzofurazan, ≥98% (elemental analysis)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
4-Fluor-7-nitrobenzofurazan, suitable for fluorescence, BioReagent, ≥98.0% (HPLC)