Merck
  • Isoprenoid geranylgeraniol: the influence on cell characteristics of endothelial progenitor cells after bisphosphonate therapy in vitro.
Isoprenoid geranylgeraniol: the influence on cell characteristics of endothelial progenitor cells after bisphosphonate therapy in vitro.
Clinical oral investigations (2015-01-16)
A M Pabst, M Krüger, T Ziebart, C Jacobs, C Walter
ZUSAMMENFASSUNG
Geranylgeraniol (GGOH) has been reported as a potential treatment option for bisphosphonate-associated osteonecrosis of the jaws (BP-ONJ). The aim of this study was to analyze the effects of GGOH on endothelial progenitor cells (EPC) after bisphosphonate treatment in vitro. EPC were incubated with different nitrogen (N-BPs: ibandronate, pamidronate, zoledronate) and a non-nitrogen-containing bisphosphonates (NN-BP: clodronate) with and without GGOH. Cell viability was measured by MTT and PrestoBlue assay. Migration ability was analyzed with a Boyden and Scratch assay. Apoptosis rates were determined by colony-forming, Tunel and ToxiLight assays. Negative effects of N-BPs on EPC were shown in all tests without GGOH. The substitution of GGOH demonstrated significantly increased cell viability (MTT: p each N-BP ≤0.004; PrestoBlue: p each N-BP <0.001) and migration ability (Boyden: p each N-BP <0.001; Scratch: p each N-BP <0.001). Concerning the apoptosis rates, increased EPC colony densities (p each N-BP ≤0.009), decreased numbers of apoptotic cells in the Tunel assay (p each N-BP <0.001), and a decreased adenylate kinase release in the ToxiLight assay (p each N-BP ≤0.03) were observed. For the clodronate-treated cells, no significant differences could be detected with or without GGOH in any assay (p each N-BP/NN-BP >0.05). GGOH cell treatment reversed the negative effects of bisphosphonates on EPC. These findings support the hypothesis that systemic or local GGOH treatment might lead to new therapeutic strategies for BP-ONJ.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Resorufin, 95%
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
Sigma-Aldrich
Zitrat-konzentrierte Lösung -Lösung, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Calcein-AM -Lösung, 4 mM in DMSO, ≥90% (HPLC), solution
Sigma-Aldrich
Zitrat-konzentrierte Lösung -Lösung, BioUltra, Molecular Biology, 1 M in H2O