Merck
  • S-equol Partially Restored Endothelial Nitric Oxide Production in Isoflavone-deficient Ovariectomized Rats.
S-equol Partially Restored Endothelial Nitric Oxide Production in Isoflavone-deficient Ovariectomized Rats.
Journal of cardiovascular pharmacology (2015-01-31)
Yoshinori Ohkura, Satoshi Obayashi, Kazuki Yamada, Mikiko Yamada, Toshiro Kubota
ZUSAMMENFASSUNG
S-equol is known as an estrogenic substance, but its ability to restore vascular endothelial function is unknown. The aim of this study was to investigate the impact of S-equol on endothelial function and intimal thickening under isoflavone- and estrogen-deficient circumstances. Twelve-week-old female Sprague-Dawley rats were bilaterally ovariectomized and assigned to one of the 3 groups: control, isoflavone-deficient (ID), or ID plus equol (n = 12, respectively). The control group received a normal diet containing isoflavones, while ID and ID plus equol groups received isoflavones-free diet. At 16th week, subcutaneous administration of S-equol (200 μg/d) started in the ID plus equol group. At 18th week, endothelial denudation of the left common carotid artery was performed in all groups, and thoracic and carotid arteries were collected at 20th week. In thoracic artery, endothelium-dependent relaxation, cyclic guanosine monophosphate levels in the tissue, and endothelial nitric oxide (NO) synthase expression and phosphorylation were significantly higher in the groups of ID plus equol and control than in the ID. The ratio of intima to media of the injured carotid artery in the control group was the lowest. Removal of dietary soy isoflavones decreased endothelium-derived NO level in ovariectomized rats. S-equol supplementation partially improved NO-related endothelial function.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Trichloressigsäure, ACS reagent, ≥99.0%
Sigma-Aldrich
Chlorotrimethylsilan, ≥98.0% (GC)
Sigma-Aldrich
Trichloressigsäure -Lösung, 6.1 N
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
Sigma-Aldrich
Zirkonium(IV)-oxid, powder, 5 μm, 99% trace metals basis
Sigma-Aldrich
Chlorotrimethylsilan, purified by redistillation, ≥99%
Sigma-Aldrich
Natriumacetat, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Trichloressigsäure, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Natriumacetat, 99.995% trace metals basis
Sigma-Aldrich
Natriumacetat, >99%, FG
Sigma-Aldrich
Zirkonium(IV)-oxid, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Trichloressigsäure, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
N-tert-Butyldimethylsilyl-N-methyltrifluoracetamid mit 1% tert-Butyldimethylchlorsilan, ≥95%
Sigma-Aldrich
Natriumacetat, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Trichloressigsäure, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloressigsäure, ≥99.0% (titration)
Sigma-Aldrich
Natriumacetat -Lösung, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
DL-Glyceraldehyd-3-phosphat -Lösung, 45-55 mg/mL in H2O
Sigma-Aldrich
Natriumacetat, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Ethylacetat, ≥99%, FCC, FG
Sigma-Aldrich
Natriumacetat, meets USP testing specifications, anhydrous
Sigma-Aldrich
Ethylacetat