Merck
  • Development of a mass-spectrometry-based lipidomics platform for the profiling of phospholipids and sphingolipids in brain tissues.
Development of a mass-spectrometry-based lipidomics platform for the profiling of phospholipids and sphingolipids in brain tissues.
Analytical and bioanalytical chemistry (2015-06-24)
Tingting Zhang, Si Chen, Xinle Liang, Hong Zhang
ZUSAMMENFASSUNG
This article describes the development of a lipidomic platform consisting of a 4000 QTRAP mass spectrometer and a self-installed sample inlet system to indentify and quantify 12 phospholipid and five sphingolipid classes from lipid-rich brain tissues of mouse, duck, and salmon. The total mass spectrometry analysis time per sample was 30 min, including 14 min for direct infusion for phospholipids and sulfatide in precursor ion scanning mode or neutral loss scanning mode, and 16 min for liquid-chromatographic separation of ceramide, sphingomyelin, monohexosylceramide, and dihexosylceramide in multiple reaction monitoring mode. The method was fully validated in terms of linearity, detectability, recovery, and repeatability, with satisfactory results for all targets. We individually quantified 307, 308, and 330 lipid species from 17 lipid subclasses, and obtained total amounts of 57.2, 61.7, and 53.1 mg/g wet brain for mouse, duck, and salmon tissues, respectively. Phosphatidylcholine, phosphatidylethanolamine, phosphatidylserine, and sphingomyelin were the major lipids in all the brain samples, whereas phosphatidylinositol occurred at a relatively higher level in the salmon sample. For phospholipids, sphingolipids, and minor lysophospholipids, differences in the identity of the molecular species, their distributions, and their relative amounts as well as the contribution of each lipid subclass to the whole polar lipidome were found. Palmitic acid (16:0), stearic acid (18:0), lignoceric acid (24:0), oleic acid (18:1), nervonic acid (24:1), arachidonic acid (20:4), and docosahexaenoic acid (22:6) were found as the main saturated, monounsaturated, and polyunsaturated fatty acids in samples from the different species, but eicosapentaenoic acid (20:5) and docosahexaenoic acid (22:6) were more abundant in the salmon brain sample. The results are in good agreement with those in previous reports obtained from the relevant tissues, providing a reliable basis that could be extended to clinical research and resource evaluation. Graphical Abstract Methodology for phospholipids and sphingolipids profiling in brain tissues.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammoniumacetat, Molecular Biology, ≥98%
Sigma-Aldrich
Stearinsäure, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
Ammoniumacetat -Lösung, Molecular Biology, 7.5 M
Sigma-Aldrich
1,3-Propandiol, 98%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Ammoniumacetat, 99.999% trace metals basis
Sigma-Aldrich
Stearinsäure, reagent grade, 95%
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Ammoniumacetat, reagent grade, ≥98%
Sigma-Aldrich
Stearinsäure, ≥95%, FCC, FG
Sigma-Aldrich
Ammoniumacetat, BioXtra, ≥98%
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, purum, ≥99.0% (TLC)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Essigsäure-12C2, 99.9 atom % 12C