Merck
Transcriptional network profile on synovial fluid T cells in psoriatic arthritis.
Clinical rheumatology (2015-07-15)
Ugo Fiocco, Veronica Martini, Benedetta Accordi, Francesco Caso, Luisa Costa, Francesca Oliviero, Anna Scanu, Monica Facco, Daniele Boso, Mariele Gatto, Mara Felicetti, Paola Frallonardo, Roberta Ramonda, Lucia Piva, Renato Zambello, Carlo Agostini, Raffaele Scarpa, Giuseppe Basso, Gianpietro Semenzato, Jean-Michel Dayer, Leonardo Punzi, Andrea Doria
ZUSAMMENFASSUNG
The objective of the study was to quantify the transcriptional profile, as the main T cell lineage-transcription factors on synovial fluid (SF) T cells, in relation to SF cytokines and T cell frequencies (%) of psoriatic arthritis (PsA) patients. Reverse phase protein array was employed to identify interleukin (IL)-23Rp19-, FOXP3- and related orphan receptor gamma T (RORγt)- protein and Janus associated tyrosine kinases 1 (JAK1), signal transducer and activator and transcription 1 (STAT1), STAT3 and STAT5 phosphoproteins in total T cell lysates from SF of PsA patients. IL-1β, IL-2, IL-6, IL-21 and interferon (INF)-γ were measured using a multiplex bead immunoassay in SF from PsA patients and peripheral blood (PB) from healthy controls (HC). Frequencies of CD4(+)CD25(-), CD4(+)CD25(high) FOXP3(+) and CD4(+)CD25(high) CD127(low) Treg, and either mean fluorescence intensity (MFI) of FOXP3(+) on CD4(+) Treg or MFI of classic IL-6 receptor (IL-6R) α expression on CD4(+)CD25(-) helper/effector T cells (Th/eff) and Treg cells, were quantified in SF of PsA patients and in PB from HC by flow cytometry (FC). In PsA SF samples, IL-2, IL-21 and IFN-γ were not detectable, whereas IL-6 and IL-1β levels were higher than in SF of non-inflammatory osteoarthritis patients. Higher levels of IL-23R-, FOXP3- and RORγt proteins and JAK1, STAT1, STAT3 and STAT5 were found in total T cells from SF of PsA patients compared with PB from HC. Direct correlations between JAK1 Y1022/Y1023 and STAT5 Y694, and STAT3 Y705 and IL6, were found in SF of PsA patients. Increased proportion of CD4(+)CD25(high) FOXP3(+) and CD4(+)CD25(high) CD127(low) Treg cells and brighter MFI of IL-6Rα were observed both on CD4(+)CD25(high)- and CD4(+)CD25(-) T cells in PsA SF. The study showed a distinctive JAK1/STAT3/STAT5 transcriptional network on T cells in the joint microenvironment, outlining the interplay of IL-6, IL-23, IL-1β and γC cytokines in the polarization and plasticity of Th17 and Treg cells, which might participate in the perpetuation of joint inflammation in PsA patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
SAFC
Glycin
Sigma-Aldrich
Glycin, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycin, ACS reagent, ≥98.5%
Sigma-Aldrich
4-(2-Aminoethyl)benzolsulfonylfluorid -hydrochlorid, ≥97.0% (HPLC)
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Glycin, 99%, FCC
Sigma-Aldrich
Glycin, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycin, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
2,2′-Bichinolin-4,4′-dicarbonsäure Dinatriumsalz Dihydrat, ≥98% (HPLC)
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis