Merck
  • Tracking and identification of antibacterial components in the essential oil of Tanacetum vulgare L. by the combination of high-performance thin-layer chromatography with direct bioautography and mass spectrometry.
Tracking and identification of antibacterial components in the essential oil of Tanacetum vulgare L. by the combination of high-performance thin-layer chromatography with direct bioautography and mass spectrometry.
Journal of chromatography. A (2015-10-27)
Ágnes M Móricz, Tim T Häbe, Andrea Böszörményi, Péter G Ott, Gertrud E Morlock
ZUSAMMENFASSUNG
Two tansy (Tanacetum vulgare L.) essential oils were obtained by steam distillation of the capitula with subsequent liquid-liquid extraction (oil 1) or with use of an auxiliary phase for the trapping of the steam components (oil 2). These oils were investigated against Bacillus subtilis F1276, B. subtilis spizizenii (DSM 618), Xanthomonas euvesicatoria, Pseudomonas syringae pv. maculicola, Ralstonia solanacearum strain GMI1000 and Aliivibrio fischeri, using the coupling of high-performance thin-layer chromatography to direct bioautography (HPTLC-DB). Using this method with the potato and tomato pathogen R. solanacearum is shown for the first time. Due to the advanced extraction process, oil 2 was richer in components and provided more inhibition zones. The main bioactive components were identified by scanning HPTLC-Direct Analysis in Real Time mass spectrometry (HPTLC-DART-MS) and solid-phase microextraction gas chromatography electron impact MS (SPME-GC-EI-MS) as cis- and trans-chrysanthenol as well as trans-chrysanthenyl acetate. cis-Chrysanthenol exhibited antibacterial effects against all tested bacteria, whereas trans-chrysanthenol inhibited B. subtilis, R. solanacearum and A. fischeri. trans-Chrysanthenyl acetate was an inhibitor for X. euvesicatoria, R. solanacearum and A. fischeri. Although HPTLC-DART-MS resulted in a comparable fragmentation, the ionization characteristics and the recorded mass spectra clearly showed that DART is a softer ionization technique than EI. It is also more affected by ambient conditions and thus prone to additional oxidation products.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumphosphat, ACS reagent, ≥99.0%
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Toluol, anhydrous, 99.8%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Kaliumphosphat, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 24 ga, Autosampler, pk of 3, notched gray hub
Sigma-Aldrich
Kaliumphosphat, ReagentPlus®
Sigma-Aldrich
Divinylbenzol, technical grade, 55%
Sigma-Aldrich
Divinylbenzol, technical grade, 80%
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Sigma-Aldrich
Kaliumphosphat, Molecular Biology, ≥98.0%
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 23 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, red hub