Merck
  • Transcriptional regulation of SLURP2, a psoriasis-associated gene, is under control of IL-22 in the skin: A special reference to the nested gene LYNX1.
Transcriptional regulation of SLURP2, a psoriasis-associated gene, is under control of IL-22 in the skin: A special reference to the nested gene LYNX1.
International immunopharmacology (2015-06-03)
Yasuhiro Moriwaki, Kiyoko Takada, Shoutaro Tsuji, Koichiro Kawashima, Hidemi Misawa
ZUSAMMENFASSUNG
A novel nicotinic acetylcholine (ACh) receptor (nAChR)-mediated transduction pathway, regulating keratinocyte function, has been elucidated in studies of secreted mammalian Ly6/urokinase plasminogen activator receptor-related protein (SLURP)-1 and -2. SLURPs are members of Ly6/neurotoxin superfamily (Ly6SF) of proteins containing the unique three-finger domain in their three-dimensional structure. Some endogenously expressed Ly6SF proteins (such as LYNX1, SLURP-1, and SLURP-2) modulate the function of nAChR, either as allosteric and/or orthosteric modulators, or as antagonists. Although the expression and functions of SLURP-1 and SLURP-2 in keratinocytes are well documented, the expression and the modes of action of LYNX1 in keratinocytes are unknown. Additionally, a particular hybrid transcript, LYNX1-SLURP2, which contains both LYNX1 and SLURP-2 sequences, with unknown function, has been reported. Furthermore, although SLURP2 is a gene strongly induced in psoriatic skin lesions, the mechanisms controlling SLURP2 expression are largely unknown. To better understand the function of nAChRs in keratinocytes, we investigated the expression profiles of LYNX1, LYNX1-SLURP-2, and SLURP-2 in keratinocytes under various inflammatory conditions. We found that keratinocytes express LYNX1 and SLURP2, but not LYNX1-SLURP2, at mRNA and protein levels. IL-22 treatment increased SLURP2 expression in keratinocytes, but this effect was completely abolished by IFN-γ. Furthermore, the IL-22-induced up-regulation of SLURP2 was completely suppressed by the inhibitor or siRNA for STAT3, a major transcriptional factor downstream of IL-22. These findings provide new insights into the nAChR-mediated regulatory mechanism of SLURP-2 expression in keratinocytes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Butanon, ACS reagent, ≥99.0%
Sigma-Aldrich
Actidion, from microbial, ≥94% (TLC)
Sigma-Aldrich
Cycloheximid -Lösung, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Cycloheximid, ≥95% (HPLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
Actidion, Biotechnology Performance Certified
Sigma-Aldrich
2-Butanon, FCC, FG
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
Sigma-Aldrich
2-Butanon, ACS reagent, ≥99.0%
Sigma-Aldrich
PD 98,059, solid
Sigma-Aldrich
S3I-201, ≥97% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human STAT3
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Stat3