Merck
  • Mass spectrometric characterisation of a condensation product between porphobilinogen and indolyl-3-acryloylglycine in urine of patients with acute intermittent porphyria.
Mass spectrometric characterisation of a condensation product between porphobilinogen and indolyl-3-acryloylglycine in urine of patients with acute intermittent porphyria.
Journal of mass spectrometry : JMS (2015-09-10)
Josep Marcos, Maria Ibañez, Rosa Ventura, Jordi Segura, Jordi To-Figueras, Oscar J Pozo
ZUSAMMENFASSUNG
We document the presence of a previously unknown species in the urine of patients with acute intermittent porphyria (AIP). The compound was fully characterised by liquid chromatography tandem mass spectrometry. Interpretation of both full spectrum acquisition and product ion spectra acquired in positive and negative ionisation modes by quadrupole time of flight MS allowed for the identification of a condensation product arising from porphobilinogen (PBG, increased in the urine of AIP patients) and indolyl-3-acryloylglycine (IAG, derived from indolylacrylic acid and present in human urine). The structure was unequivocally confirmed through comparison between the selected reaction monitoring chromatograms obtained from the urinary species and the condensation product qualitatively synthesised in the laboratory. Owing to the large amounts of both PBG and IAG in urine of AIP patients, the possible ex vivo formation of PBG-IAG in urine samples was evaluated. The product was spontaneously formed at room temperature, at 4 °C and even during storage at -20 °C when spiking a control sample with PBG. A positive correlation was found between PBG and PBG-IAG in samples collected from AIP patients. However, no correlation was found between PBG-IAG and IAG. Purified PBG-IAG did not form the characteristic chromogen after application of p-dimethylaminobenzaldehyde in HCl, thus suggesting that the current techniques used to measure PBG in urine of AIP patients based on Ehlrich's reaction do not detect this newly characterised PBG-IAG fraction.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammoniumformat, ≥99.995% trace metals basis
Sigma-Aldrich
Ammoniumformiat -Lösung, BioUltra, 10 M in H2O
Sigma-Aldrich
4-(Dimethylamino)benzaldehyd, ACS reagent, 99%
Sigma-Aldrich
4-(Dimethylamino)benzaldehyd, 98%
Sigma-Aldrich
5-Aminolevulinsäure -hydrochlorid, ≥98%
Sigma-Aldrich
5-Aminolevulinsäure -hydrochlorid, BioReagent, suitable for cell culture, powder, ≥98%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
4-(Dimethylamino)benzaldehyd, suitable for histochemical demonstration of nitro blue tetrazolium reduction in neutrophils
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
Porphobilinogen, powder
Sigma-Aldrich
4-(Dimethylamino)benzaldehyd, puriss. p.a., reag. Ph. Eur., ≥99% (perchloric acid titration)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.