Merck
  • Oxidized LDL-bound CD36 recruits an Na⁺/K⁺-ATPase-Lyn complex in macrophages that promotes atherosclerosis.
Oxidized LDL-bound CD36 recruits an Na⁺/K⁺-ATPase-Lyn complex in macrophages that promotes atherosclerosis.
Science signaling (2015-09-10)
Yiliang Chen, David J Kennedy, Devi Prasadh Ramakrishnan, Moua Yang, Wenxin Huang, Zhichuan Li, Zijian Xie, Alexandra C Chadwick, Daisy Sahoo, Roy L Silverstein
ZUSAMMENFASSUNG
One characteristic of atherosclerosis is the accumulation of lipid-laden macrophage foam cells in the arterial wall. We have previously shown that the binding of oxidized low-density lipoprotein (oxLDL) to the scavenger receptor CD36 activates the kinase Lyn, initiating a cascade that inhibits macrophage migration and is necessary for foam cell generation. We identified the plasma membrane ion transporter Na(+)/K(+)-ATPase as a key component in the macrophage oxLDL-CD36 signaling axis. Using peritoneal macrophages isolated from Atp1a1 heterozygous or Cd36-null mice, we demonstrated that CD36 recruited an Na(+)/K(+)-ATPase-Lyn complex for Lyn activation in response to oxLDL. Macrophages deficient in the α1 Na(+)/K(+)-ATPase catalytic subunit did not respond to activation of CD36, showing attenuated oxLDL uptake and foam cell formation, and oxLDL failed to inhibit migration of these macrophages. Furthermore, Apoe-null mice, which are a model of atherosclerosis, were protected from diet-induced atherosclerosis by global deletion of a single allele encoding the α1 Na(+)/K(+)-ATPase subunit or reconstitution with macrophages that lacked an allele encoding the α1 Na(+)/K(+)-ATPase subunit. These findings identify Na(+)/K(+)-ATPase as a potential target for preventing or treating atherosclerosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyaninperchlorat, BioReagent, suitable for fluorescence, ≥98.0% (TLC)
Sigma-Aldrich
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyaninperchlorat, 97%
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)