Merck
  • Modeling pulmonary alveolar microlithiasis by epithelial deletion of the Npt2b sodium phosphate cotransporter reveals putative biomarkers and strategies for treatment.
Modeling pulmonary alveolar microlithiasis by epithelial deletion of the Npt2b sodium phosphate cotransporter reveals putative biomarkers and strategies for treatment.
Science translational medicine (2015-11-13)
Atsushi Saito, Nikolaos M Nikolaidis, Hassane Amlal, Yasuaki Uehara, Jason C Gardner, Kathleen LaSance, Lori B Pitstick, James P Bridges, Kathryn A Wikenheiser-Brokamp, Dennis W McGraw, Jason C Woods, Yves Sabbagh, Susan C Schiavi, Göksel Altinişik, Marko Jakopović, Yoshikazu Inoue, Francis X McCormack
ZUSAMMENFASSUNG
Pulmonary alveolar microlithiasis (PAM) is a rare, autosomal recessive lung disorder associated with progressive accumulation of calcium phosphate microliths. Inactivating mutations in SLC34A2, which encodes the NPT2b sodium-dependent phosphate cotransporter, has been proposed as a cause of PAM. We show that epithelial deletion of Npt2b in mice results in a progressive pulmonary process characterized by diffuse alveolar microlith accumulation, radiographic opacification, restrictive physiology, inflammation, fibrosis, and an unexpected alveolar phospholipidosis. Cytokine and surfactant protein elevations in the alveolar lavage and serum of PAM mice and confirmed in serum from PAM patients identify serum MCP-1 (monocyte chemotactic protein 1) and SP-D (surfactant protein D) as potential biomarkers. Microliths introduced by adoptive transfer into the lungs of wild-type mice produce marked macrophage-rich inflammation and elevation of serum MCP-1 that peaks at 1 week and resolves at 1 month, concomitant with clearance of stones. Microliths isolated by bronchoalveolar lavage readily dissolve in EDTA, and therapeutic whole-lung EDTA lavage reduces the burden of stones in the lungs. A low-phosphate diet prevents microlith formation in young animals and reduces lung injury on the basis of reduction in serum SP-D. The burden of pulmonary calcium deposits in established PAM is also diminished within 4 weeks by a low-phosphate diet challenge. These data support a causative role for Npt2b in the pathogenesis of PAM and the use of the PAM mouse model as a preclinical platform for the development of biomarkers and therapeutic strategies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasser, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Proteasehemmer-Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Wasser, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphorsäure, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Wasser, Deionized
Sigma-Aldrich
Wasser, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Wasser, Molecular Biology, sterile filtered
Sigma-Aldrich
Phosphorsäure, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphorsäure -Lösung, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Wasser, BioPerformance Certified
Sigma-Aldrich
Phosphorsäure, BioUltra, ≥85% (T)
Sigma-Aldrich
Wasser, ACS reagent
Sigma-Aldrich
Phosphorsäure, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Wasser, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Phosphorsäure, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Wasser, PCR Reagent
Sigma-Aldrich
2,2′-Bichinolin-4,4′-dicarbonsäure Dinatriumsalz Dihydrat, ≥98% (HPLC)
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Wasser, endotoxin, free
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Wasser-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Phosphorsäure-16O4 -Lösung, 70 wt. % in D2O, 99.9 atom % 16O