Merck
Contribution of autoallergy to the pathogenesis in the NOD mice.
Autoimmunity (2015-02-25)
Radha Thyagarajan, Viqar Banday, Zhoujie Ding, Kristina Lejon
ZUSAMMENFASSUNG
The immunoglobulin isotype IgE is commonly associated with allergy. However, its involvement in autoimmune disease in general, and Type 1 diabetes (T1D) in particular, is still not completely clarified, nonetheless IgE has been observed in patients with T1D. In this article, we aimed to elucidate the contribution of IgE in the pathogenesis of the disease in a spontaneous model for T1D, i.e. the NOD mouse. We observed increased levels of IgE in splenic, lymph node and peripheral blood B cells in the NOD mice compared to the control C57BL/6 (B6) mice. No correlation was found between the IgE levels on B cells and those in the sera of these mice, indicating a B cell intrinsic property mediating IgE capture in NOD. Functionally, the B cells from NOD were similar to B6 in rescuing the IgE-mediated immune response via the low affinity receptor CD23 in a transgenic adoptive transfer system. However, the involvement of IgE in diabetes development was clearly demonstrated, as treatment with anti-IgE antibodies delayed the incidence of the diabetes in the NOD mice compared to the PBS treated group. Pancreas sections from a 13-week-old NOD revealed the presence of tertiary lymphoid structures with T cells, B cells, germinal centers and IgE suggesting the presence of autoantigen specific IgE. Our study provides an insight to the commonly overlooked immunoglobulin IgE and its potential role in autoimmunity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasser, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Wasser, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Picrylsulfonsäure -Lösung, 5 % (w/v) in H2O, BioReagent, suitable for determination of primary amines
Sigma-Aldrich
Wasser, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Ammoniumchlorid, Molecular Biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Wasser, Molecular Biology, sterile filtered
Sigma-Aldrich
Ammoniumchlorid, 99.998% trace metals basis
Sigma-Aldrich
Wasser, BioPerformance Certified
Sigma-Aldrich
Gly-Gly, ≥99% (titration)
Sigma-Aldrich
Wasser, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Ammoniumchlorid, 99.99% trace metals basis
Sigma-Aldrich
Wasser, PCR Reagent
Sigma-Aldrich
Ammoniumchlorid, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Gly-Gly, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
Gly-Gly, BioPerformance Certified, suitable for cell culture, ≥99%
Sigma-Aldrich
Wasser, endotoxin, free
Sigma-Aldrich
Gly-Gly, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Ammonium-14N-chlorid, 99.99 atom % 14N, 15N-depleted, 99% (CP)
Sigma-Aldrich
Wasser-16O, ≥99.94 atom % 16O