Merck
miR-34a inhibits cell proliferation in prostate cancer by downregulation of SIRT1 expression.
Oncology letters (2016-01-02)
Kun Duan, Yong-Chao Ge, Xue-Pei Zhang, Shu-Yi Wu, Jin-Shun Feng, Shi-Lin Chen, L I Zhang, Zhi-Hao Yuan, Chao-Hong Fu
ZUSAMMENFASSUNG
MicroRNA-34a (miR-34a) functions as a tumor suppressor gene and inhibits abnormal cell growth by regulating the expression of other genes. The role of miR-34a in regulating sirtuin 1 (SIRT1) in prostate cancer remains unclear. The objective of the present study was to investigate the biological function and molecular mechanisms of miR-34a regulation of SIRT1 in human prostate cancer samples and the human prostate cancer cell line, PC-3. Fresh prostate tissues were obtained from patients, and the miR-34a expression in prostate cancer tissues was measured using reverse transcription-quantitative polymerase chain reaction (RT-qPCR). qPCR and western blotting were performed to assess the effects of miR-34a overexpression on SIRT1 regulation in PC-3 cells, and the cell growth was assessed by Cell Counting Kit-8 (CCK-8). Flow cytometry was used to assess the cell cycle status of the cells. The miR-34a expression levels in prostate cancer tissues were significantly reduced compared with adjacent normal prostate tissues (P<0.05). SIRT1 expression levels in PC-3 cells with over-expression of miR-34a were significantly reduced compared with those in the negative control (P<0.05). The over-expression of miR-34a inhibited PC-3 cells growth and resulted in increased cell cycle arrest compared with the negative control (P<0.05). In conclusion, miR-34a inhibits the human prostate cancer cell proliferation, in part, through the downregulation of SIRT1 expression.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Luminol, 97%
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Luminol, ≥97% (HPLC)
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis