Merck
  • Effect of Ganoderma lucidum spores intervention on glucose and lipid metabolism gene expression profiles in type 2 diabetic rats.
Effect of Ganoderma lucidum spores intervention on glucose and lipid metabolism gene expression profiles in type 2 diabetic rats.
Lipids in health and disease (2015-05-23)
Fang Wang, Zhongkai Zhou, Xiaochong Ren, Yuyang Wang, Rui Yang, Jinhua Luo, Padraig Strappe
ZUSAMMENFASSUNG
The fruiting body of Ganoderma lucidum has been used as a traditional herbal medicine for many years. However, to the date, there is no detailed study for describing the effect of G. lucidum spores on oxidative stress, blood glucose level and lipid compositions in animal models of type 2 diabetic rats, in particular the effect on the gene expression profiles associated with glucose and lipid metabolisms. G. lucidum spores powder (GLSP) with a shell-broken rate >99.9 % was used. Adult male Sprague-Dawley rats were randomly divided into three groups (n = 8/group). Group 1: Normal control, normal rats with ordinary feed; Group 2: Model control, diabetic rats with ordinary feed without intervention; Group 3: GLSP, diabetic rats with ordinary feed, an intervention group utilizing GLSP of 1 g per day by oral gavages for 4 consecutive weeks. Type 2 diabetic rats were obtained by streptozocin (STZ) injection. The changes in the levels of glucose, triglycerides, total cholesterol and HDL-cholesterol in blood samples were analyzed after GLSP intervention. Meanwhile, gene expressions associated with the possible molecular mechanism of GLSP regulation were also investigated using a quantitative RT-PCR. The reduction of blood glucose level occurred within the first 2 weeks of GLSP intervention and the lipid synthesis in the diabetic rats of GLSP group was significantly decreased at 4 weeks compared to the model control group. Furthermore, it was also found that GLSP intervention greatly attenuated the level of oxidative stress in the diabetic rats. Quantitative RT-PCR analysis showed up-regulation of lipid metabolism related genes (Acox1, ACC, Insig-1 and Insig-2) and glycogen synthesis related genes (GS2 and GYG1) in GLSP group compared to model control group. Additionally, there were no significant changes in the expression of other genes, such as SREBP-1, Acly, Fas, Fads1, Gpam, Dgat1, PEPCK and G6PC1. This study might indicate that GLSP consumption could provide a beneficial effect in terms of lowering the blood glucose levels by promoting glycogen synthesis and inhibiting gluconeogenesis. Meanwhile, GLSP treatment was also associated with the improvement of blood lipid compositions through the regulation of cholesterol homeostasis in the type 2 diabetic rats.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
L-Glutathion reduziert, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathion reduziert, ≥98.0%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
L-Glutathion reduziert, BioXtra, ≥98.0%
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)