Merck
  • Cryostructuring of polymer systems. Proteinaceous wide-pore cryogels generated by the action of denaturant/reductant mixtures on bovine serum albumin in moderately frozen aqueous media.
Cryostructuring of polymer systems. Proteinaceous wide-pore cryogels generated by the action of denaturant/reductant mixtures on bovine serum albumin in moderately frozen aqueous media.
Soft matter (2015-05-27)
Ilya A Rodionov, Natalia V Grinberg, Tatiana V Burova, Valery Ya Grinberg, Vladimir I Lozinsky
ZUSAMMENFASSUNG
Freeze-thaw processing of bovine serum albumin (BSA) aqueous solutions, which contain also the additives of denaturants (urea in this case) and thiol-bearing reductants [cysteine (Cys) in this case] leads to the formation of wide-pore cryogels. The properties and porous morphology of these spongy gel matrices were demonstrated to depend on the initial concentration of all precursors and on the freezing/frozen storage temperature. The optimum conditions for preparing such BSA-based cryogels were found to be as follows: [BSA] = 3-5 g dL(-1), [urea] = 0.5-2.0 mol L(-1), [Cys] = 0.01 mol L(-1), and freezing temperatures in the range of -15 to -20 °C. The size of gross pores in thus prepared cryogels is ∼50-150 μm. The spatial network of BSA-cryogels was shown to be cross-linked chemically via interchain disulfide bridges. The significant role of hydrophobic interactions in the stabilization of 3D networks of these cryogels is inferred, as well as the supposition about the relay-race sequence mechanism of the intermolecular disulfide cross-link formation is made.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Harnstoff, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Harnstoff, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Supelco
Harnstoff, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Methylenblau, certified by the BSC
Sigma-Aldrich
Harnstoff -Lösung, BioUltra, ~8 M in H2O
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Methylenblau -Lösung, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Harnstoff, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Harnstoff, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Harnstoff, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Harnstoff, suitable for electrophoresis
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Harnstoff, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Methylenblau -Lösung, 0.05 wt. % in H2O
Sigma-Aldrich
Harnstoff, meets USP testing specifications
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Harnstoff, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
DL-Cystein, technical grade
Sigma-Aldrich
Harnstoff -Lösung, 40 % (w/v) in H2O
Sigma-Aldrich
Methylenblaulösung 1,4 % (w/v) 95 % Ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol