Merck
  • Goniothalamin enhances TRAIL-induced apoptosis in colorectal cancer cells through DR5 upregulation and cFLIP downregulation.
Goniothalamin enhances TRAIL-induced apoptosis in colorectal cancer cells through DR5 upregulation and cFLIP downregulation.
International journal of oncology (2015-10-27)
Thanet Sophonnithiprasert, Sirinun Nilwarangkoon, Yukio Nakamura, Ramida Watanapokasin
ZUSAMMENFASSUNG
The combination of TNF-related apoptosis-inducing ligand (TRAIL) and bioactive compound to enhance apoptosis in TRAIL-resistant cancer is one of cancer treatment strategies. TRAIL possesses the unique capacity to selectively induce apoptosis in cancer cells both in vitro and in vivo with little effect on normal cells. Recent studies have reported that there are many TRAIL-resistant cancers. Thus, bioactive compounds that enhance cytotoxicity of TRAIL would be potential candidates for cancer therapeutic application. This study evaluated the cytotoxic and apoptosis induction upon combined treatment of TRAIL and goniothalamin, the natural styryl-lactone compound extracted from plant Goniothalamus spp., in LoVo cells. The results showed that a combination of goniothalamin and TRAIL enhanced caspase-dependent apoptosis induction in LoVo cells via both death receptor- and mitochondrial-mediated apoptosis pathways. In addition, goniothalamin enhanced TRAIL-induced apoptosis through increased death receptor DR5 expression and decreased anti-apoptotic regulator cFLIP. Interestingly, goniothalamin increased translocation of DR5 to cell surface and consequently contributed to the enhancement of TRAIL-induced apoptosis. In conclusion, this is the first report showing the combined treatment of goniothalamin and TRAIL was able to effectively enhance TRAIL-mediated apoptosis induction in TRAIL-refractory colorectal cancer, LoVo cells. Therefore, this study may offer a strategic cancer treatment against TRAIL-resistant cancers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
TRAIL human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE and HPLC), lyophilized powder
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®
Sigma-Aldrich
TRAIL from mouse, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture
Sigma-Aldrich
TRAIL human, recombinant, expressed in NSO cells, >97% (SDS-PAGE), lyophilized powder