Merck
  • Increasing the Collision Rate of Particle Impact Electroanalysis with Magnetically Guided Pt-Decorated Iron Oxide Nanoparticles.
Increasing the Collision Rate of Particle Impact Electroanalysis with Magnetically Guided Pt-Decorated Iron Oxide Nanoparticles.
ACS nano (2015-07-15)
Donald A Robinson, Jason J Yoo, Alma D Castañeda, Brett Gu, Radhika Dasari, Richard M Crooks, Keith J Stevenson
ZUSAMMENFASSUNG
An integrated microfluidic/magnetophoretic methodology was developed for improving signal response time and detection limits for the chronoamperometric observation of discrete nanoparticle/electrode interactions by electrocatalytic amplification. The strategy relied on Pt-decorated iron oxide nanoparticles which exhibit both superparamagnetism and electrocatalytic activity for the oxidation of hydrazine. A wet chemical synthetic approach succeeded in the controlled growth of Pt on the surface of FeO/Fe3O4 core/shell nanocubes, resulting in highly uniform Pt-decorated iron oxide hybrid nanoparticles with good dispersibility in water. The unique mechanism of hybrid nanoparticle formation was investigated by electron microscopy and spectroscopic analysis of isolated nanoparticle intermediates and final products. Discrete hybrid nanoparticle collision events were detected in the presence of hydrazine, an electrochemical indicator probe, using a gold microband electrode integrated into a microfluidic channel. In contrast with related systems, the experimental nanoparticle/electrode collision rate correlates more closely with simple theoretical approximations, primarily due to the accuracy of the nanoparticle tracking analysis method used to quantify nanoparticle concentrations and diffusion coefficients. Further modification of the microfluidic device was made by applying a tightly focused magnetic field to the detection volume to attract the magnetic nanoprobes to the microband working electrode, thereby resulting in a 6-fold increase to the relative frequency of chronoamperometric signals corresponding to discrete nanoparticle impact events.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dopamin -hydrochlorid
Sigma-Aldrich
Salpetersäure, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Schwefelsäure, 99.999%
Sigma-Aldrich
Oleinsäure, technical grade, 90%
Sigma-Aldrich
Oleinsäure, ≥99% (GC)
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Oleinsäure, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumphosphat, ReagentPlus®, ≥99.0%
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Natriumphosphat, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Natriumphosphat, BioXtra, ≥99.0%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Natriumphosphat, Molecular Biology, ≥98.5% (titration)
Sigma-Aldrich
Hydrazin -Lösung, 35 wt. % in H2O
Sigma-Aldrich
Hydrazin -sulfat (Salz), ACS reagent, ≥99.0%
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Natriumphosphat, zweibasisch -Lösung, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Oleinsäure, natural, FCC
Sigma-Aldrich
Kalium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Nitrosyltetrafluorborat, 95%
Sigma-Aldrich
Oleinsäure, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Salpetersäure, ACS reagent, ≥90.0%
Sigma-Aldrich
Natriumphosphat, 99.95% trace metals basis
Sigma-Aldrich
Kaliumhydrid, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
Natriumphosphat, BioUltra, Molecular Biology, ≥99.5% (T)