Merck
  • Localized experimental bone metastasis drives osteolysis and sensory hypersensitivity at distant non-tumor-bearing sites.
Localized experimental bone metastasis drives osteolysis and sensory hypersensitivity at distant non-tumor-bearing sites.
Breast cancer research and treatment (2015-07-26)
Dareen M Abdelaziz, Laura S Stone, Svetlana V Komarova
ZUSAMMENFASSUNG
Patients with breast cancer metastasis to bone suffer from inadequate pain relief. Animal models provide increased understanding of cancer-induced bone and sensory alterations. The objective of this study was to investigate the measures of pain at distant non-tumor-bearing sites in animals with localized bone metastasis. Immunocompetent BALB/c mice are injected intra-tibially with murine mammary carcinoma cells (4T1) or saline, and the sensitivity to mechanical and thermal stimuli in the contralateral paw was examined. In addition to previously demonstrated development of osteolysis and hypersensitivity to mechanical and thermal stimuli in the cancer-injected tibia, these animals exhibited an increase in sensory hypersensitivity in the contralateral limb. No bone lesions were evident on radiographs of the contralateral limbs. Histomorphometry detected decreased bone volume per tissue volume and increased osteoclast number in the contralateral tibia and vertebral bones of cancer-bearing animals. Neuroplasticity was examined by immunofluorescence for calcitonin gene-related peptide (CGRP) in sensory neurons and glial fibrillary acidic protein (GFAP) in lumbar spinal cords. CGRP-immunoreactivity and GFAP-immunoreactivity were significantly elevated both ipsilateral and contralateral in tumor-bearing animals. The anti-inflammatory and osteolysis-targeting drug rapamycin reduced hypersensitivity to mechanical and cold stimuli, attenuated GFAP over-expression, and lowered osteoclast number. The osteoclast-targeting drug pamidronate reduced sensitivity to cold and protected against bone loss. Localized bone cancer drives hypersensitivity, bone remodeling, and sensory neuron plasticity at sites distant from the primary tumor area. Drugs targeting these mechanisms may be useful in the treatment of pain distant from the primary tumor site.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
L-Lysin -monohydrochlorid, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Natriumbicarbonat, BioXtra, 99.5-100.5%
Sigma-Aldrich
Poly(ethylenglykol), average mol wt 400
Sigma-Aldrich
L-Lysin -monohydrochlorid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
L-Lysin -monohydrochlorid, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumbicarbonat-12C, 13C-depleted, 99.9 atom % 12C
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis