Merck
Phenolics from Caesalpinia ferrea Mart.: antioxidant, cytotoxic and hypolipidemic activity.
Die Pharmazie (2015-09-19)
M A Nawwar, S A Hussein, A M El-Mousallami, A N Hashim, M A Mousa, M H Hetta, M A Hamed, V Werner, A Becker, B Haertel, U Lindequist
ZUSAMMENFASSUNG
Nine phenolics were isolated from the aqueous ethanol extract of the leaves of Caesalpinia ferrea. The isolates were characterized for the first time from that plant. The structures of all isolates (1-9) were elucidated by conventional methods, spectroscopic analysis, including 1 D and 2D NMR, and by HR-ESIMS as well. The antioxidant capacities using the ORAC method and the cytotoxic activity using the neutral red assay (NRU) for that extract and three major isolates have been evaluated. In addition, the hypolipidemic activity (in vitro and in vivo) of the extract has been assessed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid-d6, 99.9 atom % D
Sigma-Aldrich
Dimethylsulfoxid-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Dimethylsulfoxid-d6, 99.5 atom % D
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Neutralrot-Lösung (0,33 %), 3.3 g/L in DPBS, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Etoposid, synthetic, 95.0-105.0%, powder
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Dimethylsulfoxid-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethylsulfoxid-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Dimethylsulfoxid-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Neutralrot, Dye content ≥90 %
Sigma-Aldrich
DL-3-Hydroxy-3-methylglutaryl-Coenzym-A Natriumsalz, ≥90% (HPLC)
Sigma-Aldrich
Neutralrot, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethylsulfoxid-d6, "Special HOH", ≥99.9 atom % D
Sigma-Aldrich
Neutralrot, certified by the BSC
Sigma-Aldrich
Neutralrot -Lösung
Sigma-Aldrich
Dimethylsulfoxid-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Dimethylsulfoxid-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethylsulfoxid-d6, 99.9 atom % D
Sigma-Aldrich
Dimethylsulfoxid-d6, "100%", 99.96 atom % D