Merck
  • Fabrication and characterization of a highly sensitive hydroquinone chemical sensor based on iron-doped ZnO nanorods.
Fabrication and characterization of a highly sensitive hydroquinone chemical sensor based on iron-doped ZnO nanorods.
Dalton transactions (Cambridge, England : 2003) (2015-11-26)
Ahmad Umar, Ali Al-Hajry, Rafiq Ahmad, S G Ansari, Mohammed Sultan Al-Assiri, Hamed Algarni
ZUSAMMENFASSUNG
Herein, we report the development of a simple and highly sensitive hydroquinone (HQ) chemical sensor based on an electrochemically activated iron-doped (Fe-doped) zinc oxide nanorod (ZnO NR) modified screen-printed electrode (SPE). The Fe-doped ZnO NRs were prepared using a hydrothermal process and their morphological, crystal, compositional and optical properties were characterized in detail. The detailed characterizations showed that the NRs are densely grown, well-crystalline and possess a wurtzite hexagonal phase. The fabricated HQ electrochemical sensor exhibited high sensitivity of 18.60 μA mM(-1) cm(-2) and a very low experimental detection limit of 0.51 μM. Our results demonstrate that simply prepared doped ZnO nanomaterials are promising candidates to fabricate highly sensitive electrochemical sensors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ammoniumhydroxid -Lösung, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Ammoniumhydroxid -Lösung, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Hexamethylentetramin, ACS reagent, ≥99.0%
Sigma-Aldrich
Hydrochinon, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydrochinon, ReagentPlus®, 99%
Sigma-Aldrich
Hexamethylentetramin, ReagentPlus®, 99%
Sigma-Aldrich
Hexamethylentetramin, puriss. p.a., reag. Ph. Eur., ≥99.5% (calc. to the dried substance)
Sigma-Aldrich
Hexamethylentetramin, meets analytical specification of Ph. Eur., BP, 99-100.5% (calc. to the dried substance)
Sigma-Aldrich
Hydrochinon, meets USP testing specifications