Merck
  • Photocatalytic degradation of pentachlorophenol by visible light Ν-F-TiO₂ in the presence of oxalate ions: optimization, modeling, and scavenging studies.
Photocatalytic degradation of pentachlorophenol by visible light Ν-F-TiO₂ in the presence of oxalate ions: optimization, modeling, and scavenging studies.
Environmental science and pollution research international (2015-01-22)
M Antonopoulou, I Konstantinou
ZUSAMMENFASSUNG
The efficiency of heterogeneous photocatalysis using N-F-TiO2 as photocatalyst to degrade a priority pollutant, pentachlorophenol (PCP), in the presence of oxalates (OA) was investigated in detail. Response surface methodology was used to optimize the effect of three variables (catalyst concentration, OA/PCP ratio, and pH) on the photocatalytic degradation of pentachlorophenol. A quadratic model was established as a functional relationship between three independent variables and the degradation efficiency of PCP. The results of model fitting and statistical analysis demonstrated that the pH played a key role in the degradation of PCP. Within the studied experimental ranges, the optimum conditions for maximum PCP degradation efficiency (97.5 %) were: catalyst concentration 600 mg L(-1), OA/PCP ratio 2, and pH 10. The contribution of HO(·), O2 (·-), and e(-) produced during the photocatalytic treatment was investigated with the addition of scavengers. The photocatalytic degradation was essentially proceeded through an oxidative mechanism at both acid and alkaline pH values by HO(.) and O2 (·-) radicals attack. It was found that O2 (·-) were the major reactive species involved in PCP degradation in pH 4 and HO(·) in pH 10.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Oxalsäure, 98%
Sigma-Aldrich
Oxalsäure, ReagentPlus®, ≥99%
Sigma-Aldrich
Oxalsäure, purified grade, 99.999% trace metals basis
Sigma-Aldrich
Oxalsäure, puriss. p.a., anhydrous, ≥99.0% (RT)
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
4-Chlor-DL-Phenylalanin