Merck
  • Control of Histamine-Producing Bacteria and Histamine Formation in Fish Muscle by Trisodium Phosphate.
Control of Histamine-Producing Bacteria and Histamine Formation in Fish Muscle by Trisodium Phosphate.
Journal of food science (2015-04-30)
Kristin Bjornsdottir-Butler, David P Green, Greg E Bolton, Patricia D McClellan-Green
ZUSAMMENFASSUNG
Scombrotoxin fish poisoning remains the primary cause of seafood poisoning outbreaks despite preventive guidelines. The purpose of this study was to investigate the use of pH for the control of growth and histamine formation by histamine-producing bacteria in fish muscle. We examined pH effects on growth and histamine formation in tuna fish infusion broth and in inoculated tuna and mahi-mahi fish muscle. Histamine production was significantly less for all bacterial strains at pH 8.5 compared to pH 5.5 in tuna fish infusion broth with no significant difference in growth. Elevated pH due to phosphate treatment of fish muscle tissues significantly reduced histamine formation with no effect on the growth of histamine-producing bacteria. This study revealed that phosphate treatment of mahi-mahi and tuna fish muscle resulted in significantly lower histamine production over 4 d of storage at 10 °C. Phosphate treatment of fish muscle may serve as a secondary barrier in addition to FDA recommended time and temperature controls for reducing public health concerns of scombrotoxin fish poisoning.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Histamin, ≥97.0%
Sigma-Aldrich
Poly(tetrafluorethylen), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Natriumhydroxid, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Poly(tetrafluorethylen), powder, 200 μm particle size
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Poly(tetrafluorethylen), powder, >40 μm particle size
Sigma-Aldrich
Poly(tetrafluorethylen), beads
Sigma-Aldrich
Poly(tetrafluorethylen), powder, 35 μm particle size
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Poly(tetrafluorethylen), powder (free-flowing), ≤12 μm particle size
Sigma-Aldrich
Poly(tetrafluorethylen), powder, ≥350 μm particle size