Merck
Donkey Milk for Manufacture of Novel Functional Fermented Beverages.
Journal of food science (2015-05-13)
Annamaria Perna, Immacolata Intaglietta, Amalia Simonetti, Emilio Gambacorta
ZUSAMMENFASSUNG
The aim of this work was to investigate on the functional features of a donkey milk probiotic berevage as a novel food. Particularly, it was to study the decrease of lactose content and the antioxidant activity of standard yogurt (YC) and probiotic yogurt (YP; Lactobacillus acidophilus, Lactobacillus casei) from donkey milk during the storage up to 30 d at 4 ºC. The evolution of lactose content using enzymatic-spectrophotometric kits was analyzed. Antioxidant activity of yogurt was measured using 2,2'-azino-bis-3-ethylbenzothiazoline-6-sulfonic acid (ABTS), ferric reducing antioxidant power (FRAP), and thiol assays. Parallel consumer sensory studies were carried out as consumer test in order to gain information about the impact of these novel fermented beverages on sensory perceptions. The statistical analysis has shown significant effect of studied factors. The results showed that the lactose content gradually decreased during storage in both yogurts, reaching values of 2.36% and 2.10% in YC and YP, respectively, at 30 d (P < 0.05). During storage of both yogurt types, the antioxidant activity increased, but YP showed a higher antioxidant activity than YC. The results suggest that the antioxidant activity of yogurt samples was affected by cultures of lactic acid bacteria (LAB). We conclude that the fermented donkey milk could be configured as health and nutraceutical food, which aims to meet nutritional requirements of certain consumers groups with lactose or cow milk protein intolerance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoesäure), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Natriumphosphat, 96%
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoesäure), ReagentPlus®, 99%
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Supelco
2,4,6-Tri-(2-pyridyl)-s-triazin, ≥98%
Sigma-Aldrich
Natriumacetat, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Eisen(III)-chlorid, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Natriumacetat, 99.995% trace metals basis
Sigma-Aldrich
Salzsäure -Lösung, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Natriumacetat, >99%, FG
Sigma-Aldrich
Kaliumpersulfat, 99.99% trace metals basis
Sigma-Aldrich
Eisen(III)-chlorid, ≥99.9% trace metals basis
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Natriumacetat, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
Natriumacetat -Lösung, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Natriumacetat, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG