Merck
  • Dynamic interplay of multidrug transporters with TolC for isoprenol tolerance in Escherichia coli.
Dynamic interplay of multidrug transporters with TolC for isoprenol tolerance in Escherichia coli.
Scientific reports (2015-11-14)
Chonglong Wang, Liyang Yang, Asad Ali Shah, Eui-Sung Choi, Seon-Won Kim
ZUSAMMENFASSUNG
Engineering of efflux pumps is a promising way to improve host's tolerance to biofuels such as medium-chain alcohols (CmOHs); however, this strategy is restricted by poor understanding of the efflux pumps engaged in extrusion of solvents. In this study, several Escherichia coli mutants of multidrug transporters were evaluated for isoprenol tolerance. Susceptible phenotypes were observed in the mutants with individual deletion of six transporters, AcrD, EmrAB, MacAB, MdtBC, MdtJI and YdiM, whereas inactivation of AcrAB transporter resulted in an improved tolerance to isoprenol and other CmOHs. AcrAB is the major transporter forming tripartite transperiplasmic complex with outer membrane channel TolC for direct extrusion of toxic molecules in E. coli. The AcrAB inactivation enables to enhance TolC availability for the multidrug transporters associated with extrusion of CmOHs and increase the tolerance to CmOHs including isoprenol. It is assumed that outer membrane channel TolC plays an important role in extrusion of isoprenol and other CmOHs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
1-Pentanol, ReagentPlus®, ≥99%
Sigma-Aldrich
1-Hexanol, reagent grade, 98%
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
3-Methyl-1-Butanol, reagent grade, 98%
Sigma-Aldrich
Isoamylalkohol, ≥98%, FG
Sigma-Aldrich
1-Pentanol, ACS reagent, ≥99%
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Sigma-Aldrich
3-Methyl-1-Butanol, anhydrous, ≥99%
Sigma-Aldrich
3-Methyl-1-Butanol, ACS reagent, ≥98.5%
Sigma-Aldrich
1-Hexanol, anhydrous, ≥99%
Sigma-Aldrich
1-Pentanol, ≥99%, FG
Sigma-Aldrich
Isoamylalkohol, natural, ≥98%, FG
Sigma-Aldrich
Butylalkohol, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
1-Pentanol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
3-Methylbutanol, BioReagent, ≥98.5%, Molecular Biology
Sigma-Aldrich
L-Thyroxin Natriumsalz Pentahydrat, ≥98% (HPLC), powder
Sigma-Aldrich
3-Methylbutanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Hexylalkohol, natural, ≥98%, FCC, FG
Sigma-Aldrich
Hexylalkohol, FCC, FG
Sigma-Aldrich
L-Thyroxin Natriumsalz Pentahydrat, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
1-Hexanol, ReagentPlus®, ≥99.5% (GC)
Sigma-Aldrich
1-Butanol, BioRenewable, ACS reagent, ≥99.4%