Merck
  • Measurement of breakthrough volumes of volatile chemical warfare agents on a poly(2,6-diphenylphenylene oxide)-based adsorbent and application to thermal desorption-gas chromatography/mass spectrometric analysis.
Measurement of breakthrough volumes of volatile chemical warfare agents on a poly(2,6-diphenylphenylene oxide)-based adsorbent and application to thermal desorption-gas chromatography/mass spectrometric analysis.
Journal of chromatography. A (2015-08-05)
Mieko Kanamori-Kataoka, Yasuo Seto
ZUSAMMENFASSUNG
To establish adequate on-site solvent trapping of volatile chemical warfare agents (CWAs) from air samples, we measured the breakthrough volumes of CWAs on three adsorbent resins by an elution technique using direct electron ionization mass spectrometry. The trapping characteristics of Tenax(®) TA were better than those of Tenax(®) GR and Carboxen(®) 1016. The latter two adsorbents showed non-reproducible breakthrough behavior and low VX recovery. The specific breakthrough values were more than 44 (sarin) L/g Tenax(®) TA resin at 20°C. Logarithmic values of specific breakthrough volume for four nerve agents (sarin, soman, tabun, and VX) showed a nearly linear correlation with the reciprocals of their boiling points, but the data point of sulfur mustard deviated from this linear curve. Next, we developed a method to determine volatile CWAs in ambient air by thermal desorption-gas chromatography (TD-GC/MS). CWA solutions that were spiked into the Tenax TA(®) adsorbent tubes were analyzed by a two-stage TD-GC/MS using a Tenax(®) TA-packed cold trap tube. Linear calibration curves for CWAs retained in the resin tubes were obtained in the range between 0.2pL and 100pL for sarin, soman, tabun, cyclohexylsarin, and sulfur mustard; and between 2pL and 100pL for VX and Russian VX. We also examined the stability of CWAs in Tenax(®) TA tubes purged with either dry or 50% relative humidity air under storage conditions at room temperature or 4°C. More than 80% sarin, soman, tabun, cyclohexylsarin, and sulfur mustard were recovered from the tubes within 2 weeks. In contrast, the recoveries of VX and Russian VX drastically reduced with storage time at room temperature, resulting in a drop to 10-30% after 2 weeks. Moreover, we examined the trapping efficiency of Tenax TA(®) adsorbent tubes for vaporized CWA samples (100mL) prepared in a 500mL gas sampling cylinder. In the concentration range of 0.2-2.5mg/m(3), >50% of sarin, soman, tabun, cyclohexylsarin, and HD were recovered, whereas <1% of VX and Russian VX were recovered in the same concentration range. The results indicate that CWA vapors, with the exception of VX and Russian VX, can be measured by an on-site collection procedure using the Tenax(®) TA resin tubes, followed by a subsequent TD-GC/MS analysis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 24 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 23 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, plain gray hub
Supelco
ORBO Carboxen-564 Röhrchen, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
Supelco
ORBO 78 HBr auf Carboxen® 564 speziell gereinigt (20/45), 400/200 mg, W,W,W separators, O.D. × L 6 mm × 110 mm, pkg of 25 ea
Supelco
Carboxen®-1010 PLOT Kapillar-GC-Säule, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
Carboxen® Adsorptionsmittel, matrix Carboxen® 1000, 60-80 mesh, bottle of 10 g
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, black hub
Supelco
Carboxen®-1006 PLOT Kapillar-GC-Säule, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, black hub
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 85 μm CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, light blue hub
Supelco
Carboxen®-1010 PLOT Kapillar-GC-Säule, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 85 μm CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, light blue hub
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 85 μm CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, light blue hub
Supelco
ORBO-93 Sorbensröhrchen, ORBO tube I.D. × L 6 mm × 95 mm, Bed A 180 mg, Bed B 90 mg, 60/80 mesh, pkg of 25 ea
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, black hub
Supelco
Carboxen®-1006 PLOT Kapillar-GC-Säule, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Supelco
Tenax® TA / Carboxen® 1018, glass TD tube, preconditioned, O.D. × L 1/4 in. × 3 1/2 in., Sealed with (Swagelok® End-Fittings), pkg of 10 ea
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, black hub
Supelco
Carboxen® Adsorptionsmittel, matrix Carboxen® 1003, 40-60 mesh, bottle of 10 g
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, Metal Alloy (2cm), needle size 23 ga, Autosampler, pk of 1, plain gray hub
Supelco
Carboxen® 572 for Tobacco Smoke, Adsorbent Bed Wt: 300 mg, cartridge size 3 mL, pkg of 50 ea
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, Metal Alloy (1cm), needle size 23 ga, Autosampler, pk of, plain gray hub
Supelco
Carboxen® Adsorptionsmittel, matrix Carboxen® 569, 20-45 mesh, bottle of 10 g
Supelco
Carboxen® 1000 Reversibles SPE-Röhrchen, 200 mg/mL, pk 30
Supelco
Carboxen® Adsorptionsmittel, matrix Carboxen® 572, 20-45 mesh, bottle of 10 g
Supelco
Carboxen® Adsorptionsmittel, matrix Carboxen® 1000, 40-60 mesh, bottle of 50 g
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 85 μm CAR/PDMS, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, light blue hub