Merck
  • Photocatalytic removal of 2-nitrophenol using silver and sulfur co-doped TiO₂under natural solar light.
Photocatalytic removal of 2-nitrophenol using silver and sulfur co-doped TiO₂under natural solar light.
Water science and technology : a journal of the International Association on Water Pollution Research (2015-07-24)
Mehrzad Feilizadeh, Amin Delparish, S Toufigh Bararpour, Hamed Abedini Najafabadi, S Mohammad Esmaeil Zakeri, Manouchehr Vossoughi
ZUSAMMENFASSUNG
To overcome the drawback of poor solar light utilization brought about by the narrow photoresponse range of TiO₂, a silver and sulfur co-doped TiO₂was synthesized. Using the prepared catalyst, solar photocatalytic degradation of 2-nitrophenol (2-NP) by a TiO₂-based catalyst was studied for the first time. Effects of the co-doping on the structural, optical and morphological properties of the synthesized nanoparticles were investigated by different characterization methods: X-ray diffraction, N2 adsorption-desorption measurements, scanning electron microscopy, transmission electron microscopy, energy-dispersive X-ray spectroscopy, UV-visible diffuse reflectance spectroscopy and Fourier transform infrared spectroscopy. Solar experiments showed that the co-doping with silver and sulfur significantly increased the photocatalytic activity. In various initial concentrations of 2-NP more than 99% of the contaminant was decomposed by Ag-S/TiO₂in less than 150 minutes, while the degradation efficiency was much less in the presence of bare TiO₂. Kinetic studies suggested that solar photocatalytic degradation of 2-NP is consistent with the Langmuir-Hinshelwood model. The rate constant of the reaction and adsorption constant of the modified photocatalyst were found to be 2.4 and 4.1 times larger than that of bare TiO₂, respectively.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Titan(IV)isopropoxid, 97%
Sigma-Aldrich
Silbernitrat, 99.9999% trace metals basis
Sigma-Aldrich
Thioharnstoff, ACS reagent, ≥99.0%
Sigma-Aldrich
Titan(IV)isopropoxid, 99.999% trace metals basis
Sigma-Aldrich
Titan(IV)isopropoxid, ≥97.0%
Sigma-Aldrich
Thioharnstoff, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Silbernitrat auf Kieselgel, extent of labeling: ~10 wt. % loading, +230 mesh
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Sigma-Aldrich
Silbernitrat -Lösung, 2.5 % (w/v) AgNO3 in H2O
Sigma-Aldrich
Silbernitrat, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
Silbernitrat, ≥99.999% trace metals basis
Sigma-Aldrich
Titan(IV)isopropoxid, packaged for use in deposition systems
Sigma-Aldrich
Silbernitrat, BioXtra, >99% (titration)
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, purum, ≥99.0% (TLC)
Sigma-Aldrich
Essigsäure-12C2, 99.9 atom % 12C