Merck
  • Lipoxin B₄ promotes the resolution of allergic inflammation in the upper and lower airways of mice.
Lipoxin B₄ promotes the resolution of allergic inflammation in the upper and lower airways of mice.
Mucosal immunology (2014-12-04)
L Karra, O Haworth, R Priluck, B D Levy, F Levi-Schaffer
ZUSAMMENFASSUNG
Chronic mucosal inflammation is the hallmark of important and common airway diseases, such as allergic rhinitis (AR) and asthma. Lipoxin A4 (LXA4) is an endogenous pro-resolving mediator for mucosal inflammation that decreases allergic and asthmatic responses. Lipoxin B4 (LXB4) is a structurally distinct member of the lipoxin family that signals in a manner distinct from LXA4. LXB4 is generated by mucosal tissues, but its actions in allergic inflammation are unknown. Here, we used murine models of AR and asthma to investigate LXB4's activity in mucosal inflammation. In the upper airway, LXB4 significantly decreased nasal mucosal leukocytes and degranulation of mast cells (MCs) and eosinophils. In the lower airway, LXB4 significantly decreased airway inflammation, mucus metaplasia, and hyper-responsiveness. Inhibition of MC degranulation in vivo by LXB4 was more potent than dexamethasone, and these agents displayed unique profiles for cytokine regulation; however, their overall anti-inflammatory actions were comparable. LXB4 decreased eotaxin-dependent eosinophil chemotaxis, IgE-mediated MC degranulation, and expression of type 2 cytokine receptors. Together, these findings indicate that LXB4 carries cell type selective and mucosal protective actions that broaden the lipoxin family's therapeutic potential for upper and lower airway catabasis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasser, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Wasser, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Dexamethason, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Wasser, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Wasser, Molecular Biology, sterile filtered
Sigma-Aldrich
Dexamethason, ≥98% (HPLC), powder
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Hämatoxylin
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Wasser, BioPerformance Certified
Sigma-Aldrich
Formaldehyd -Lösung, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Hämatoxylin, certified by the BSC
Sigma-Aldrich
Dexamethason, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Wasser, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Wasser, PCR Reagent
Sigma-Aldrich
Dexamethason, meets USP testing specifications
Sigma-Aldrich
Aluminium-Kaliumsulfat Dodecahydrat, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Wasser, endotoxin, free
Sigma-Aldrich
Periodsäure, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Aluminium-Kaliumsulfat Dodecahydrat, BioXtra, ≥98.0%
Sigma-Aldrich
Periodsäure, 99.999% trace metals basis
Sigma-Aldrich
Formaldehyd-12C -Lösung, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Wasser-16O, ≥99.94 atom % 16O