Merck
  • Human mesenchymal stem cells as a novel platform for simultaneous evaluation of cytotoxicity and genotoxicity of pharmaceuticals.
Human mesenchymal stem cells as a novel platform for simultaneous evaluation of cytotoxicity and genotoxicity of pharmaceuticals.
Mutagenesis (2015-01-02)
Shikha Sharma, Vijayalakshmi Venkatesan, Balakrishna Murthy Prakhya, Ramesh Bhonde
ZUSAMMENFASSUNG
The in vitro micronucleus test is a well-known test for the screening of genotoxic compounds. However until now, most studies have been performed on either human peripheral lymphocytes or established cancer cell lines. This study provides human mesenchymal stem cells as an alternative to the conventional micronucleus test. We grew umbilical cord mesenchymal stem cells (UC-MSCs) on coverslips eliminating the cumbersome technique involving hypotonic treatment, fixation and preparing smears required for suspension culture (lymphocytes). The background frequency of nuclear blebs and micronuclei in UC-MSCs was found to be 7±5, in lymphocytes 16±3.5 and 9±3 and that for A549 cell line was 65±5 and 15±5 per 1000 cells, respectively, suggesting differences in the repair mechanism of normal and cancer cell lines. We inspected the cytotoxic and genotoxic effects of two known mutagens, mitomycin-C and hydrogen peroxide (H2O2), on UC-MSCs, lymphocytes and A549 cells. Treatment with mitomycin-C and H2O2 demonstrated drastic differences in the degree of cytotoxicity and genotoxicity suggesting a constitutional difference between normal and cancer cells. In addition we tested two solvents, dimethyl sulfoxide (DMSO) and ethanol, and two drugs, metformin and rapamycin. DMSO above 1% was found to be cytotoxic and genotoxic, whereas ethanol at same concentration was neither cytotoxic nor genotoxic indicating the minimal non-toxic level of the solvents. This study thus offers UC-MSCs as a better substitute to peripheral lymphocytes and cancer cell lines for high throughput screening of compounds and reducing the animal studies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
3-Isobutyl-1-methylxanthin, ≥99% (HPLC), powder
Sigma-Aldrich
Dexamethason, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
L-Ascorbinsäure, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
3-Isobutyl-1-methylxanthin, BioUltra, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
L-Ascorbinsäure, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbinsäure, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbinsäure, 99%
Sigma-Aldrich
L-Ascorbinsäure, reagent grade, crystalline
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dexamethason, ≥98% (HPLC), powder
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
L-Ascorbinsäure, ACS reagent, ≥99%
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
L-Ascorbinsäure, meets USP testing specifications
Sigma-Aldrich
β-Glycerin-phosphat Dinatriumsalz Pentahydrat, ≥98.0% (NT)
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
L-Ascorbinsäure, reagent grade
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 34.5-36.5%
Sigma-Aldrich
Dexamethason, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
L-Ascorbinsäure, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbinsäure, FCC, FG