Merck
Thoracic Duct Narrowing-Innovative Technique Restraining Weight Gain in Rats.
Obesity surgery (2015-10-09)
Barak Rosenzweig, Iris Barshack, Dror Harats, Aviv Shaish
ZUSAMMENFASSUNG
The lymphatic system is responsible for the absorption of fats from the digestive system, conveying 60-70 % of ingested fat to the blood stream. From the anatomical point of view, all the lymphatic drainage from the lower half of the body converges in the abdomen to enter the thoracic duct. This experimental study aim was to study the result of thoracic duct narrowing (TDN), an innovative surgical technique, on weight gain restrain in high-fat diet-fed rats. Forty-seven rats were allocated into three groups: thoracic duct narrowing ("S"-surgery), sham operation ("CS"-control surgery), and no surgery ("C"-control). All rats were fed with high-fat, cholesterol-rich diet. Food consumption and metabolic syndrome parameters including weight gain, plasma lipids and glucose, blood pressure, and viscera weight and histopathology were analyzed. Thoracic duct narrowing was proved simple and safe surgical procedure in the rat model. TDN induced weight gain restrain, associated with mild hepatic steatosis compared to moderate-severe hepatic steatosis in control groups. Splenomegaly and splenic fatty histiocytes were shown in the treated animals. TDN improved several parameters of the metabolic syndrome in high-fat diet-fed rats. TDN carries the potential of innovative obesity treatment using the lymphatic route of lipid absorption.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Hämatoxylin
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Hämatoxylin, certified by the BSC
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)