Merck
  • Amplified solid-state electrochemiluminescence detection of cholesterol in near-infrared range based on CdTe quantum dots decorated multiwalled carbon nanotubes@reduced graphene oxide nanoribbons.
Amplified solid-state electrochemiluminescence detection of cholesterol in near-infrared range based on CdTe quantum dots decorated multiwalled carbon nanotubes@reduced graphene oxide nanoribbons.
Biosensors & bioelectronics (2015-06-19)
Juan Huan, Qian Liu, Airong Fei, Jing Qian, Xiaoya Dong, Baijing Qiu, Hanping Mao, Kun Wang
ZUSAMMENFASSUNG
An amplified solid-state electrochemiluminescence (ECL) biosensor for detection of cholesterol in near-infrared (NIR) range was constructed based on CdTe quantum dots (QDs) decorated multiwalled carbon nanotubes@reduced graphene nanoribbons (CdTe-MWCNTs@rGONRs), which were prepared by electrostatic interactions. The CdTe QDs decorated on the MWCNTs@rGONRs resulted in the amplified ECL intensity by ~4.5 fold and decreased onset potential by ~100 mV. By immobilization of the cholesterol oxidase (ChOx) and NIR CdTe-MWCNTs@rGONRs on the electrode surface, a solid-state ECL biosensor for cholesterol detection was constructed. When cholesterol was added to the detection solution, the immobilized ChOx catalyzed the oxidation of cholesterol to generate H2O2, which could be used as the co-reactant in the ECL system of CdTe-MWCNTs@rGONRs. The as-prepared biosensor exhibited good performance for cholesterol detection including good reproducibility, selectivity, and acceptable linear range from 1 μM to 1mM with a relative low detection limit of 0.33 μM (S/N=3). The biosensor was successfully applied to the determination of cholesterol in biological fluid and food sample, which would open a new possibility for development of solid-state ECL biosensors with NIR emitters.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Natriumborhydrid, powder, ≥98.0%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Natriumborhydrid, ReagentPlus®, 99%
Sigma-Aldrich
1-Methoxy-2-propyl-acetat, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Isopropylalkohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Natriumborhydrid, granular, 99.99% trace metals basis
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Natriumborhydrid, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Tellur, powder, −200 mesh, 99.8% trace metals basis
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Natriumborhydrid, granular, 10-40 mesh, 98%
Sigma-Aldrich
Natriumborhydrid -Lösung, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Tellur, powder, −30 mesh, 99.997% trace metals basis
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Tellur, 99.999% trace metals basis
Sigma-Aldrich
Natriumborhydrid, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Natriumborhydrid -Lösung, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Tellur, shot, ≤2 mm, 99.999% trace metals basis
Sigma-Aldrich
Tellur, granular, −5-+50 mesh, 99.99% trace metals basis
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)