Merck
  • Persistent organic pollutants and promoter hypermethylation of the O(6)-methylguanine-DNA methyltransferase gene.
Persistent organic pollutants and promoter hypermethylation of the O(6)-methylguanine-DNA methyltransferase gene.
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals (2015-01-15)
Soo Yeun Park, Ki-Su Kim, Yu-Mi Lee, Mi-Jin Kim, David R Jacobs, Miquel Porta, Dong-Sun Kim, Duk-Hee Lee
ZUSAMMENFASSUNG
Promoter hypermethylation of the O(6)-methylguanine-DNA methyltransferase (MGMT) DNA repair gene is important during carcinogenesis. We explored whether organochlorine pesticides (OCPs) and polychlorinated biphenyls (PCBs), were associated with hypermethylation of the MGMT gene promoter in peripheral leukocytes among 368 Koreans without cancer. Hypermethylation decreased as OCPs increased (Ptrend = 0.02), while PCB concentrations showed an inverted U-shaped association (Pquadratic < 0.01). The prevalence of MGMT promoter hypermethylation was highest within the 2nd quintile of the PCB summary score (28.4%), while it was only 2.7% in the upper 10% score. Chronic exposure to these chemicals may affect methylation of the MGMT promoter, with possibly non-monotonic dose response relationships.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)