Merck
  • In vitro activity of Melaleuca alternifolia (tea tree) oil on filamentous fungi and toxicity to human cells.
In vitro activity of Melaleuca alternifolia (tea tree) oil on filamentous fungi and toxicity to human cells.
Medical mycology (2015-01-30)
Diane C Homeyer, Carlos J Sanchez, Katrin Mende, Miriam L Beckius, Clinton K Murray, Joseph C Wenke, Kevin S Akers
ZUSAMMENFASSUNG
Invasive fungal wound infections (IFIs) are increasingly reported in trauma patients and cause considerable morbidity and mortality despite standard of care treatment in trauma centers by experienced medical personnel. Topical agents such as oil of melaleuca, also known as tea tree oil (TTO), have been proposed for adjunctive treatment of IFIs. We evaluated the activity of TTO against filamentous fungi associated with IFIs by testing 13 clinical isolates representing nine species via time-kill assay with seven concentrations of TTO (100%, 75%, 50%, 25%, 10%, 5%, and 1%). To ascertain the safety of topical application to wounds, cell viability assays were performed in vitro using human fibroblasts, keratinocytes, osteoblasts, and umbilical vein endothelial cells with 10 concentrations of TTO (75%, 50%, 25%, 10%, 5%, and 10-fold serial dilutions from 1 to 0.0001%) at five time points (5, 15, 30, 60, and 180 min). Compatibility of TTO with explanted porcine tissues was also assessed with eight concentrations of TTO (100%, 75%, 50%, 25%, 10%, 5%, 1%, and 0.1%) at the time points used for cellular assays and at 24 h. The time-kill studies showed that fungicidal activity was variable between isolates. The effect of TTO on cell viability was primarily concentration dependent with significant cytotoxicity at concentrations of ≥ 10% and ≥ 50% for cells lines and whole tissue, respectively. Our findings demonstrate that TTO possesses antifungal activity against filamentous fungi associated with IFIs; furthermore that negligible effects on whole tissues, in contrast to individual cells, were observed following exposure to TTO. Collectively, these findings indicate a potential use of TTO as topical treatment of IFIs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Hydrocortison, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Hydrocortison, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Hydrocortison, ≥98% (HPLC)
SAFC
L-Glutamin
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
Hydrocortison, meets USP testing specifications
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®