Merck
  • MiR-34a suppresses amphiregulin and tumor metastatic potential of head and neck squamous cell carcinoma (HNSCC).
MiR-34a suppresses amphiregulin and tumor metastatic potential of head and neck squamous cell carcinoma (HNSCC).
Oncotarget (2015-03-13)
Jiali Zhang, Yu Wang, Xinming Chen, Yi Zhou, Fangyan Jiang, Jirong Chen, Li Wang, Wen-Feng Zhang
ZUSAMMENFASSUNG
MiR-34a is a well-known tumor metastasis inhibitor, but only a few target genes involved in metastasis have been identified. In HNSCC, the role of miR-34a in metastasis has not been fully elaborated, and the target gene of miR-34a is still blind. Here we addressed that, the relative lower expression of miR-34a is associated with HNSCC lymphatic metastasis. HNSCC metastasis was found to be strongly suppressed in vitro and in vivo by over-expressing miR-34a. In order to screen the possible target genes of miR-34a in HNSCC, a microarray-based differential mRNA profiling mediated by miR-34a over-expression was performed, and AREG was identified as a pivotal target. We demonstrated that the mRNA and protein levels of AREG were greatly reduced when forcing miR-34a expression. The correlation between AREG mRNA levels and HNSCC metastatic phenotype was also significant in HNSCC tissues (p < 0.01). Moreover, the results of luciferase assay provided the further evidence that miR-34a degraded AREG mRNA through targeting the 3'-UTR site. Restoration of AREG expression partially rescued miR-34a-mediated cell invasion defects in vivo and in vitro. Additionally, Over-expressing miR-34a greatly reduced EGFR and uPA, which were reversed by re-expression of AREG. Taken together, these findings indicate that miR-34a targets AREG, and is essential in inhibition of HNSCC metastasis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Deoxycholsäure Natriumsalz, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Deoxycholsäure Natriumsalz, ≥97% (titration)
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
SAFC
Natriumchlorid -Lösung, 5 M
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
SAFC
Deoxycholsäure Natriumsalz
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)