Merck
Influence of cholesterol on liposome stability and on in vitro drug release.
Drug delivery and translational research (2015-03-20)
Maria-Lucia Briuglia, Chiara Rotella, Amber McFarlane, Dimitrios A Lamprou
ZUSAMMENFASSUNG
Cholesterol plays a strategic role in liposome composition; however, the quantity used to achieve an appropriate formulation has not been yet clarified. Therefore, by screening arrangement of lipids and cholesterol ratio, the main aim of this study is to investigate the most suitable amount of cholesterol in lipids in order to prepare stable and controlled drug release vehicles. For the preparation of liposomes, DMPC, DPPC and DSPC phospholipids were used and combined with different molar ratios of cholesterol (e.g. 100, 80-20, 70-30, 60-40 and 50-50%). Stability studies were conducted by storing the formulations at 37 and 50 °C for 30 days and by analysing them by AFM, DLS and FT-IR. By detecting the two most stable formulations from the stability results, drug encapsulation and in vitro release studies in PBS were performed by encapsulating atenolol and quinine. The release results were validated using a simulation model to ensure the reliability and suitable interpretation of the data. The generated model showed a good correlation between the prediction and the in vitro obtained results. By using 70:30% ratio (known in literature as 2:1), it is possible to reach the most stable formulation to guarantee a controlled and reproducible release for drugs with different physicochemical characteristics and pharmaceutical applications.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Chinin, 90%
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
Sigma-Aldrich
Chinin, suitable for fluorescence, anhydrous, ≥98.0% (dried material, NT)
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)