Merck
The Dimerization State of the Mammalian High Mobility Group Protein AT-Hook 2 (HMGA2).
PloS one (2015-06-27)
Lorraine Frost, Maria A M Baez, Christopher Harrilal, Alyssa Garabedian, Francisco Fernandez-Lima, Fenfei Leng
ZUSAMMENFASSUNG
The mammalian high mobility group protein AT-hook 2 (HMGA2) is a chromosomal architectural transcription factor involved in cell transformation and oncogenesis. It consists of three positively charged "AT-hooks" and a negatively charged C-terminus. Sequence analyses, circular dichroism experiments, and gel-filtration studies showed that HMGA2, in the native state, does not have a defined secondary or tertiary structure. Surprisingly, using combined approaches of 1-Ethyl-3-(3-dimethylaminopropyl)carbodiimide hydrochloride (EDC) chemical cross-linking, analytical ultracentrifugation, fluorescence resonance energy transfer (FRET), and mass spectrometry, we discovered that HMGA2 is capable of self-associating into homodimers in aqueous buffer solution. Our results showed that electrostatic interactions between the positively charged "AT-hooks" and the negatively charged C-terminus greatly contribute to the homodimer formation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tris(2-carboxyethyl)phosphin -hydrochlorid, powder
Supelco
Tris(2-carboxyethyl)phosphin -hydrochlorid -Lösung, 0.5 M, pH 7.0(aqueous solution; pH was adjusted with ammonium hydroxide)
Sigma-Aldrich
1,2-Dichlorethan, anhydrous, 99.8%
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimid -hydrochlorid, commercial grade, powder
Sigma-Aldrich
Tris(2-carboxyethyl)phosphin -hydrochlorid, BioUltra, ≥98% (NMR)
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimid -hydrochlorid, ≥99.0% (AT)
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimid -hydrochlorid, crystalline
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimid -hydrochlorid, BioXtra
Sigma-Aldrich
Tris(2-carboxyethyl)phosphin -hydrochlorid, BioUltra, suitable for electrophoresis, SDS-PAGE tested