Merck
  • Immobilization of Cyclooxygenase-2 on Silica Gel Microspheres: Optimization and Characterization.
Immobilization of Cyclooxygenase-2 on Silica Gel Microspheres: Optimization and Characterization.
Molecules (Basel, Switzerland) (2015-11-12)
Qian Shi, Junhui Chen, Yanlong Wang, Zhaoyong Li, Xianguo Li, Chengjun Sun, Li Zheng
ZUSAMMENFASSUNG
In this study, immobilized COX-2 was successfully constructed through glutaraldehyde-mediated covalent coupling on functional silica gel microspheres. The optimum conditions, properties, and morphological characteristics of the immobilized COX-2 were investigated. The optimal immobilization process was as follows: about 0.02 g of aminated silica gel microspheres was activated by 0.25% GA solution for 6 h and mixed with 5 U of free recombinant COX-2 solution. Then, the mixture was shaken for 8 h at 20 °C. Results showed that the immobilized COX-2 produced by this method exhibited excellent biocatalytic activity, equivalent to that of free COX-2 under the test conditions employed. The best biocatalytic activity of immobilized COX-2 appeared at pH 8.0 and still maintained at about 84% (RSD < 7.39%, n = 3) at pH 10.0. For temperature tolerance, immobilized COX-2 exhibited its maximum biocatalytic activity at 40 °C and about 68% (RSD < 6.99%, n = 3) of the activity was maintained at 60 °C. The immobilized COX-2 retained over 85% (RSD < 7.26%, n = 3) of its initial biocatalytic activity after five cycles, and after 10 days storage, the catalytic activity of immobilized COX-2 still maintained at about 95% (RSD < 3.08%, n = 3). These characteristics ensured the convenient use of the immobilized COX-2 and reduced its production cost.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ammoniumpersulfat, ACS reagent, ≥98.0%
Sigma-Aldrich
Ammoniumpersulfat, Molecular Biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Ammoniumpersulfat, reagent grade, 98%
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Arachidonsäure, >95.0% (GC)
Sigma-Aldrich
Arachidonsäure, from non-animal source, ≥98.5% (GC)
Sigma-Aldrich
Ammoniumpersulfat, BioUltra, ≥98.0% (RT)
Sigma-Aldrich
Ammoniumpersulfat, BioXtra, ≥98.0%
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Cyclooxygenase 2 human, ≥70% (SDS-PAGE), recombinant, expressed in insect cells, aqueous solution, ≥8000 units/mg protein
Sigma-Aldrich
Adenosine 5′-phosphosulfate sodium salt, ≥85%
Sigma-Aldrich
Ammoniumpersulfat, free-flowing, Redi-Dri, reagent grade, 98%