Merck
  • Uses of 3-(2-Bromoacetyl)-2H-chromen-2-one in the Synthesis of Heterocyclic Compounds Incorporating Coumarin: Synthesis, Characterization and Cytotoxicity.
Uses of 3-(2-Bromoacetyl)-2H-chromen-2-one in the Synthesis of Heterocyclic Compounds Incorporating Coumarin: Synthesis, Characterization and Cytotoxicity.
Molecules (Basel, Switzerland) (2015-06-26)
Rafat M Mohareb, Nadia Y MegallyAbdo
ZUSAMMENFASSUNG
In this work, 3-bromoacetylcoumarin was used as the key starting material for the synthesis of pyran, pyridine, thiophene, thiazole and pyrazole derivatives through its reaction with different reagents. The structures of the newly synthesized compounds were confirmed on the basis of their spectral data and elemental analyses. All of the synthesized compounds were screened for their in vitro anticancer activity against six human cancer cell lines, namely: human gastric cancer (NUGC), human colon cancer (DLD1), human liver cancer (HA22T and HEPG2), nasopharyngeal carcinoma (HONE1), human breast cancer (MCF) and normal fibroblast cells (WI38). The IC50 values (the sample concentration that produces 50% reduction in cell growth) in nanomolars (nM)) showed most of the compounds exhibited significant cytotoxic effect. Among these derivatives, compound 6d showed almost equipotent cytotoxic activity against NUGC (IC50 = 29 nM) compared to the standard CHS 828 (IC50 = 25 nM).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
SAFC
L-Glutamin
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Sulforhodamin B Mononatriumsalz, Dye content 75 %
Sigma-Aldrich
Sulforhodamin B Natriumsalz, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Cholesterylhemisuccinat Tris-salz, anionic detergent
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
Sulforhodamin B Natriumsalz, Technical grade
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®