Merck
  • Examining concentrations and molecular weights of thiols in microorganism cultures and in Churchill River (Manitoba) using a fluorescent-labeling method coupled to asymmetrical flow field-flow fractionation.
Examining concentrations and molecular weights of thiols in microorganism cultures and in Churchill River (Manitoba) using a fluorescent-labeling method coupled to asymmetrical flow field-flow fractionation.
Analytical and bioanalytical chemistry (2015-03-17)
Vaughn Mangal, Celine Guéguen
ZUSAMMENFASSUNG
In this study, molecular weights of thiols from four laboratory cultures (Scenedesmus obliquus, Chlorella vulgaris, Euglena gracilis, and Attheya septentrionalis) and the Churchill River (Manitoba) were assessed using a fluorescent-labeling method such as monobromotrimethylammoniobimane (qBBr) and asymmetrical flow field-flow fractionation (AF4) coupled to a fluorescence detector. Concentrations of thiols in extracellular fractions ranged from 6.39 ± 3.39 to 39.2 ± 7.43 μmol g(-1), and intracellular concentrations ranged from 11.5 ± 4.52 to 41.0 ± 4.1 μmol g(-1). In addition, molecular weights (MW) of intracellular thiol ranged from 493 ± 24 to 946 ± 12 Da whereas extracellular thiol MWs varied from 443 ± 36 to 810 ± 174 Da. The novel method of combining AF4 to an on-line fluorometer and the incorporation of the thiol tag provided information regarding thiol concentration and composition of controlled and natural systems. Furthermore, the proposed methods allow for the simultaneous measurement of thiol and DOM MWs produced by microorganisms. By assessing characteristics of naturally produced thiols and lab-grown thiols, information regarding heavy metal complexation can be determined.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Rhodamin B, ≥95% (HPLC)
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Rhodamin B, suitable for fluorescence
Sigma-Aldrich
Salzsäure -Lösung, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
Natriumhydroxid, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Sigma-Aldrich
3-Ethyl-2,4-pentandion, Tautormermischung, 98%
Sigma-Aldrich
Rhodamin B, Dye content 90 %
Sigma-Aldrich
Natriumhydroxid-16O -Lösung, 20 wt. % in H216O, 99.9 atom % 16O