Merck
Genetic variants of dental plaque Methanobrevibacter oralis.
European journal of clinical microbiology & infectious diseases : official publication of the European Society of Clinical Microbiology (2015-01-31)
H T T Huynh, V D Nkamga, M Drancourt, G Aboudharam
ZUSAMMENFASSUNG
Methanobrevibacter oralis is the major methanogenic archaea found in the oral cavity. It has been implicated in periodontitis, including the severe form. It is unknown whether certain M. oralis genetic variants are associated with severe periodontitis. Here, we developed multispacer sequence typing (MST) as a sequencing-based genotyping method for the assessment of M. oralis. The sequencing of four intergenic spacers from a collection of 17 dental plaque M. oralis isolates obtained from seven individuals revealed 482 genetic polymorphisms, including 401 single nucleotide polymorphisms (83.2 %), 55 deletions (11.4 %) and 26 insertions (5.4 %). Concatenation of the four spacers yielded nine genotypes, which were clustered into six groups with an index of discrimination of 0.919. One periodontitis patient may have harboured up to three genetic variants of M. oralis, revealing the previously unknown diversity of this archaea. MST will allow for the study of the dynamics of M. oralis populations, including inter-individual transmission and any correlations with the severity of periodontitis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesiumchlorid, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesiumchlorid -Lösung, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesiumchlorid -Lösung, 0.1 M
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~0.025 M in H2O