Merck
  • Isotope-specific and amino acid-specific heavy atom substitutions alter barrier crossing in human purine nucleoside phosphorylase.
Isotope-specific and amino acid-specific heavy atom substitutions alter barrier crossing in human purine nucleoside phosphorylase.
Proceedings of the National Academy of Sciences of the United States of America (2015-08-26)
Javier Suarez, Vern L Schramm
ZUSAMMENFASSUNG
Computational chemistry predicts that atomic motions on the femtosecond timescale are coupled to transition-state formation (barrier-crossing) in human purine nucleoside phosphorylase (PNP). The prediction is experimentally supported by slowed catalytic site chemistry in isotopically labeled PNP (13C, 15N, and 2H). However, other explanations are possible, including altered volume or bond polarization from carbon-deuterium bonds or propagation of the femtosecond bond motions into slower (nanoseconds to milliseconds) motions of the larger protein architecture to alter catalytic site chemistry. We address these possibilities by analysis of chemistry rates in isotope-specific labeled PNPs. Catalytic site chemistry was slowed for both [2H]PNP and [13C, 15N]PNP in proportion to their altered protein masses. Secondary effects emanating from carbon-deuterium bond properties can therefore be eliminated. Heavy-enzyme mass effects were probed for local or global contributions to catalytic site chemistry by generating [15N, 2H]His8-PNP. Of the eight His per subunit, three participate in contacts to the bound reactants and five are remote from the catalytic sites. [15N, 2H]His8-PNP had reduced catalytic site chemistry larger than proportional to the enzymatic mass difference. Altered barrier crossing when only His are heavy supports local catalytic site femtosecond perturbations coupled to transition-state formation. Isotope-specific and amino acid specific labels extend the use of heavy enzyme methods to distinguish global from local isotope effects.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Jodacetamid, BioUltra
Sigma-Aldrich
Jodacetamid, Single use vial of 56 mg
Sigma-Aldrich
Jodacetamid, ≥99% (NMR), crystalline
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Ammoniumchlorid, Molecular Biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Ammoniumchlorid, 99.998% trace metals basis
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Deuteriumoxid, filtered, 99.8 atom % D
Sigma-Aldrich
Ammoniumchlorid, 99.99% trace metals basis
Sigma-Aldrich
Deuteriumoxid, 99.8 atom % D
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
SAFC
Jodacetamid
Sigma-Aldrich
Ammoniumchlorid, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Deuteriumoxid, 70 atom % D
Sigma-Aldrich
DL-Histidin, ≥99% (TLC)
Sigma-Aldrich
Deuteriumoxid, 60 atom % D
Sigma-Aldrich
Ammonium-14N-chlorid, 99.99 atom % 14N, 15N-depleted, 99% (CP)
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D