Merck
  • Development of Monoclonal Antibody LpMab-10 Recognizing Non-glycosylated PLAG1/2 Domain Including Thr34 of Human Podoplanin.
Development of Monoclonal Antibody LpMab-10 Recognizing Non-glycosylated PLAG1/2 Domain Including Thr34 of Human Podoplanin.
Monoclonal antibodies in immunodiagnosis and immunotherapy (2015-10-23)
Satoshi Ogasawara, Hiroharu Oki, Mika K Kaneko, Yasukazu Hozumi, Xing Liu, Ryusuke Honma, Yuki Fujii, Takuro Nakamura, Kaoru Goto, Michiaki Takagi, Yukinari Kato
ZUSAMMENFASSUNG
Podoplanin (PDPN) is a type-I transmembrane sialoglycoprotein that possesses a platelet aggregation-stimulating (PLAG) domain in the N-terminus. PLAG domain includes three tandem repeats of eight amino acids: PLAG1, PLAG2, and PLAG3. Among the three PLAG domains, O-glycan on Thr52 of PLAG3 is critical for binding with C-type lectin-like receptor-2 (CLEC-2) and is essential for platelet-aggregating activity of PDPN. In contrast, the glycosylation of Thr34 of PLAG1 of human PDPN remains to be clarified. Herein, we developed and characterized a novel anti-PDPN monoclonal antibody, LpMab-10, which targets PLAG1/2 domain. LpMab-10 detects endogenous PDPN of cancer cells and normal cells independently of glycosylation. The minimum epitope of LpMab-10 was identified as Glu33-Gly45 of PDPN using Western blot and flow cytometry. The Thr34 of PLAG1 is critical for LpMab-10 recognition, and O-glycan is not included in LpMab-10 epitope, indicating that Thr34 of PLAG1 is not O-glycosylated. In immunocytochemical and immunohistochemical analyses, LpMab-10 strongly detected PDPN-expressing tumor cells. By using monoclonal antibodies against different Ser/Thr, including epitopes of PDPN, it becomes possible to determine whether Ser/Thr residues of PDPN are O-glycosylated.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
Aluminium-Kaliumsulfat Dodecahydrat, ACS reagent, ≥98%
Sigma-Aldrich
Thymidin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
SAFC
L-Glutamin
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Hypoxanthin, ≥99.0%
Sigma-Aldrich
Hämatoxylin
Sigma-Aldrich
Thymidin, ≥99%
Sigma-Aldrich
Hypoxanthin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
SAFC
Glycin