Merck
  • M1 and M2 macrophage proteolytic and angiogenic profile analysis in atherosclerotic patients reveals a distinctive profile in type 2 diabetes.
M1 and M2 macrophage proteolytic and angiogenic profile analysis in atherosclerotic patients reveals a distinctive profile in type 2 diabetes.
Diabetes & vascular disease research (2015-05-15)
Charlotte Roma-Lavisse, Madjid Tagzirt, Christophe Zawadzki, Rodrigo Lorenzi, André Vincentelli, Stephan Haulon, Francis Juthier, Antoine Rauch, Delphine Corseaux, Bart Staels, Brigitte Jude, Eric Van Belle, Sophie Susen, Giulia Chinetti-Gbaguidi, Annabelle Dupont
ZUSAMMENFASSUNG
This study aimed to investigate atherosclerotic mediators' expression levels in M1 and M2 macrophages and to focus on the influence of diabetes on M1/M2 profiles. Macrophages from 36 atherosclerotic patients (19 diabetics and 17 non-diabetics) were cultured with interleukin-1β (IL-1β) or IL-4 to induce M1 or M2 phenotype, respectively. The atherosclerotic mediators' expression was evaluated by quantitative reverse transcription-polymerase chain reaction (RT-PCR). The results showed that M1 and M2 macrophages differentially expressed mediators involved in proteolysis and angiogenesis processes. The proteolytic balance (matrix metalloproteinase-9 (MMP-9)/tissue inhibitor of metalloproteinase-1 (TIMP-1), MMP-9/plasminogen activator inhibitor-1 (PAI-1) and MMP-9/tissue factor pathway inhibitor-2 (TFPI-2) ratios) was higher in M1 versus M2, whereas M2 macrophages presented higher angiogenesis properties (increased vascular endothelial growth factor/TFPI-2 and tissue factor/TFPI-2 ratios). Moreover, M1 macrophages from diabetics displayed more important proangiogenic and proteolytic activities than non-diabetics. This study reveals that M1 and M2 macrophages could differentially modulate major atherosclerosis-related pathological processes. Moreover, M1 macrophages from diabetics display a deleterious phenotype that could explain the higher plaque vulnerability observed in these subjects.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Laurinsäure, ≥98%, FCC, FG
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Nitrotetrazolium-Blauchlorid, ≥90.0% (HPLC)
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99% (GC)
Sigma-Aldrich
Laurinsäure, 98%
Sigma-Aldrich
D-(+)-Mannose, ≥99% (GC), wood
Sigma-Aldrich
Vaskulärer endothelialer Wachstumsfaktor human, VEGF, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Nitrotetrazolium-Blauchlorid, powder, electrophoresis grade
Sigma-Aldrich
Laurinsäure, natural, ≥98%, FCC, FG
Sigma-Aldrich
Dodecansäure, ≥99% (GC/titration)
Sigma-Aldrich
IL-1 beta human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
VEGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture