Merck
Anti-CD20/CD3 T cell-dependent bispecific antibody for the treatment of B cell malignancies.
Science translational medicine (2015-05-15)
Liping L Sun, Diego Ellerman, Mary Mathieu, Maria Hristopoulos, Xiaocheng Chen, Yijin Li, Xiaojie Yan, Robyn Clark, Arthur Reyes, Eric Stefanich, Elaine Mai, Judy Young, Clarissa Johnson, Mahrukh Huseni, Xinhua Wang, Yvonne Chen, Peiyin Wang, Hong Wang, Noel Dybdal, Yu-Waye Chu, Nicholas Chiorazzi, Justin M Scheer, Teemu Junttila, Klara Totpal, Mark S Dennis, Allen J Ebens
ZUSAMMENFASSUNG
Bispecific antibodies and antibody fragments in various formats have been explored as a means to recruit cytolytic T cells to kill tumor cells. Encouraging clinical data have been reported with molecules such as the anti-CD19/CD3 bispecific T cell engager (BiTE) blinatumomab. However, the clinical use of many reported T cell-recruiting bispecific modalities is limited by liabilities including unfavorable pharmacokinetics, potential immunogenicity, and manufacturing challenges. We describe a B cell-targeting anti-CD20/CD3 T cell-dependent bispecific antibody (CD20-TDB), which is a full-length, humanized immunoglobulin G1 molecule with near-native antibody architecture constructed using "knobs-into-holes" technology. CD20-TDB is highly active in killing CD20-expressing B cells, including primary patient leukemia and lymphoma cells both in vitro and in vivo. In cynomolgus monkeys, CD20-TDB potently depletes B cells in peripheral blood and lymphoid tissues at a single dose of 1 mg/kg while demonstrating pharmacokinetic properties similar to those of conventional monoclonal antibodies. CD20-TDB also exhibits activity in vitro and in vivo in the presence of competing CD20-targeting antibodies. These data provide rationale for the clinical testing of CD20-TDB for the treatment of CD20-expressing B cell malignancies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, ≥97.5% (HPLC)