Merck
Fortified versus incurred residues: extraction of furazolidinone metabolite from prawn.
Analytical and bioanalytical chemistry (2015-04-12)
Lesley Johnston, Meg Croft, John Murby
ZUSAMMENFASSUNG
Most methods reported in the literature for determination of nitrofuran metabolites use the same extraction and derivatisation conditions to hydrolyse and extract the protein-bound residues from animal tissue. While undertaking certification of reference materials for nitrofuran metabolites in freeze-dried prawn, it was found that these conditions are satisfactory for recovery of spiked residues; however, extraction efficiencies of incurred furazolidinone could be substantially increased by further optimisation of the extraction conditions. The availability of a suitable certified reference material allows laboratories to ensure that their method is optimised for incurred residues.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
2-Nitrobenzaldehyd, 98%
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Semicarbazid -hydrochlorid, ≥99%
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, purum, ≥99.0% (TLC)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Essigsäure-12C2, 99.9 atom % 12C