Merck
TAZ promotes temozolomide resistance by upregulating MCL-1 in human glioma cells.
Biochemical and biophysical research communications (2015-06-06)
Tian Tian, Aimin Li, Hong Lu, Ran Luo, Mingzhi Zhang, Zhaoming Li
ZUSAMMENFASSUNG
Temozolomide is a novel cytotoxic agent currently used as first-line chemotherapy for glioblastoma multiforme (GBM). However, intrinsic or acquired chemoresistance to temozolomide remains the greatest obstacle to the successful treatment of human GBM. The principal mechanism responsible for this resistance is largely unknown. In the present study, we showed that expression of transcriptional co-activator with PDZ-binding motif (TAZ) in glioma cells correlated with temozolomide chemoresistance in human glioma cells. Overexpression of TAZ promoted temozolomide resistance in U-87MG cells, whereas knockdown of TAZ expression sensitized temozolomide-resistant U-251MG cells to temozolomide. Further, TAZ inhibits temozolomide induced apoptosis via upregulation of MCL-1 (myeloid cell leukemia 1) and high expression of TAZ predicts a poor prognosis for GBM patients. In conclusion, our results suggest that TAZ had a critical role in the resistance to temozolomide in glioma cells, and it may provide a promising target for improving the therapeutic outcome of temozolomide-resistant gliomas.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Temozolomid, ≥98% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
3-(Biphenyl-4-yl)-5-(4-tert-butylphenyl)-4-phenyl-4H-1,2,4-Triazol, 97%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®