Merck
Iron Fluoroanions and Their Clusters by Electrospray Ionization of a Fluorinating Ionic Liquid.
Journal of the American Society for Mass Spectrometry (2015-05-09)
Christopher A Zarzana, Gary S Groenewold, Michael T Benson, James Delmore, Tetsuya Tsuda, Rika Hagiwara
ZUSAMMENFASSUNG
Metal fluoroanions are of significant interest for fundamental structure and reactivity studies and for making isotope ratio measurements that are free from isobaric overlap. Iron fluoroanions [FeF(4)](-) and [FeF(3)](-) were generated by electrospray ionization of solutions of Fe(III) and Fe(II) with the fluorinating ionic liquid 1-ethyl-3-methylimidazolium fluorohydrogenate [EMIm](+)[F(HF)(2.3)](-). Solutions containing Fe(III) salts produce predominately uncomplexed [FeF(4)](-) in the negative ion spectrum, as do solutions containing salts of Fe(II). This behavior contrasts with that of solutions of FeCl(3) and FeCl(2) (without [EMIm](+)[F(HF)(2.3)](-)) that preserve the solution-phase oxidation state by producing the gas-phase halide complexes [FeCl(4)](-) and [FeCl(3)](-), respectively. Thus, the electrospray-[EMIm](+)[F(HF)(2.3)](-) process is oxidative with respect to Fe(II). The positive ion spectra of Fe with [EMIm](+)[F(HF)(2.3)](-) displays cluster ions having the general formula [EMIm](+) (n+1)[FeF(4)](-) n, and DFT calculations predict stable complexes, both of which substantiate the conclusion that [FeF(4)](-) is present in solution stabilized by the imidazolium cation. The negative ion ESI mass spectrum of the Fe-ionic liquid solution has a very low background in the region of the [FeF(4)](-) complex, and isotope ratios measured for both [FeF(4)](-) and adventitious [SiF(5)](-) produced values in close agreement with theoretical values; this suggests that very wide isotope ratio measurements should be attainable with good accuracy and precision when the ion formation scheme is implemented on a dedicated isotope ratio mass spectrometer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis