Merck
  • Molecular Characterisation of the Haemagglutinin Glycan-Binding Specificity of Egg-Adapted Vaccine Strains of the Pandemic 2009 H1N1 Swine Influenza A Virus.
Molecular Characterisation of the Haemagglutinin Glycan-Binding Specificity of Egg-Adapted Vaccine Strains of the Pandemic 2009 H1N1 Swine Influenza A Virus.
Molecules (Basel, Switzerland) (2015-06-10)
Vincenzo Carbone, Elena K Schneider, Steve Rockman, Mark Baker, Johnny X Huang, Chi Ong, Matthew A Cooper, Elizabeth Yuriev, Jian Li, Tony Velkov
ZUSAMMENFASSUNG
The haemagglutinin (HA) glycan binding selectivity of H1N1 influenza viruses is an important determinant for the host range of the virus and egg-adaption during vaccine production. This study integrates glycan binding data with structure-recognition models to examine the impact of the K123N, D225G and Q226R mutations (as seen in the HA of vaccine strains of the pandemic 2009 H1N1 swine influenza A virus). The glycan-binding selectivity of three A/California/07/09 vaccine production strains, and purified recombinant A/California/07/09 HAs harboring these mutations was examined via a solid-phase ELISA assay. Wild-type A/California/07/09 recombinant HA bound specifically to α2,6-linked sialyl-glycans, with no affinity for the α2,3-linked sialyl-glycans in the array. In contrast, the vaccine virus strains and recombinant HA harboring the Q226R HA mutation displayed a comparable pattern of highly specific binding to α2,3-linked sialyl-glycans, with a negligible affinity for α2,6-linked sialyl-glycans. The D225G A/California/07/09 recombinant HA displayed an enhanced binding affinity for both α2,6- and α2,3-linked sialyl-glycans in the array. Notably its α2,6-glycan affinity was generally higher compared to its α2,3-glycan affinity, which may explain why the double mutant was not naturally selected during egg-adaption of the virus. The K123N mutation which introduces a glycosylation site proximal to the receptor binding site, did not impact the α2,3/α2,6 glycan selectivity, however, it lowered the overall glycan binding affinity of the HA; suggesting glycosylation may interfere with receptor binding. Docking models and 'per residues' scoring were employed to provide a structure-recognition rational for the experimental glycan binding data. Collectively, the glycan binding data inform future vaccine design strategies to introduce the D225G or Q226R amino acid substitutions into recombinant H1N1 viruses.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Imidazol, ReagentPlus®, 99%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Imidazol, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Imidazol, Molecular Biology, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Imidazol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazol-Pufferlösung, BioUltra, 1 M in H2O
Sigma-Aldrich
Imidazol, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Imidazol, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazol, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Imidazol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
6′-Sialyllactose Natriumsalz, from bovine milk or colostrum, ≥97% (HPLC)
Sigma-Aldrich
N-Acetylneuraminosyl-D-lactose, from bovine milk or colostrum, ≥95% (HPLC)
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Imidazol, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Imidazol, Molecular Biology, ≥99% (titration), free-flowing, Redi-Dri