Merck
  • Effects of polyphenol-rich plant products from grape or hop as feed supplements on iron, zinc and copper status in piglets.
Effects of polyphenol-rich plant products from grape or hop as feed supplements on iron, zinc and copper status in piglets.
Archives of animal nutrition (2015-06-23)
Anja Fiesel, Melanie Ehrmann, Denise K Geßner, Erika Most, Klaus Eder
ZUSAMMENFASSUNG
Polyphenol-rich plant products as feed supplements have been shown to exert beneficial effects on feed efficiency in piglets. However, tannins as components of polyphenol-rich plant products are able to reduce the absorption of various trace elements. The present study investigated the effect of two polyphenol-rich dietary supplements, grape seed and grape marc meal extract (GME) and spent hops (SH), on iron (Fe), zinc (Zn) and copper (Cu) status in piglets supplied adequately with those trace elements. A trial with three groups of piglets which received a Control diet or the same diet supplemented with either 1% GME or 1% SH over a period of 4 weeks was performed. Concentrations of Fe, Zn and Cu in plasma, total iron binding capacity and saturation of transferrin in plasma did not differ between the three groups. Piglets fed the diet supplemented with SH showed no differences in the concentrations of Fe, Zn and Cu in the liver in comparison to the Control group. Piglets fed the diets supplemented with GME showed slightly lower concentrations of Zn and Cu in the liver than Control piglets (p < 0.05); however, concentrations of both elements remained in the physiological range. Overall, this study shows that the polyphenol-rich plant products GME and SH had marginal effect on the status of Fe, Zn and Cu in piglets.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
Gallussäure, 97.5-102.5% (titration)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 34.5-36.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder