Merck
  • Expression and characterization of three Aurora kinase C splice variants found in human oocytes.
Expression and characterization of three Aurora kinase C splice variants found in human oocytes.
Molecular human reproduction (2015-05-23)
Jessica E Fellmeth, Derek Gordon, Christian E Robins, Richard T Scott, Nathan R Treff, Karen Schindler
ZUSAMMENFASSUNG
Chromosome segregation is an extensively choreographed process yet errors still occur frequently in female meiosis, leading to implantation failure, miscarriage or offspring with developmental disorders. Aurora kinase C (AURKC) is a component of the chromosome passenger complex and is highly expressed in gametes. Studies in mouse oocytes indicate that AURKC is required to regulate chromosome segregation during meiosis I; however, little is known about the functional significance of AURKC in human oocytes. Three splice variants of AURKC exist in testis tissue. To determine which splice variants human oocytes express, we performed quantitative real-time PCR using single oocytes and found expression of all three variants. To evaluate the functional differences between the variants, we created green fluorescent protein-tagged constructs of each variant to express in oocytes from Aurkc(-/-) mice. By quantifying metaphase chromosome alignment, cell cycle progression, phosphorylation of INCENP and microtubule attachments to kinetochores, we found that AURKC_v1 was the most capable of the variants at supporting metaphase I chromosome segregation. AURKC_v3 localized to chromosomes properly and supported cell cycle progression to metaphase II, but its inability to correct erroneous microtubule attachments to kinetochores meant that chromosome segregation was not as accurate compared with the other two variants. Finally, when we expressed the three variants simultaneously, error correction was more robust than when they were expressed on their own. Therefore, oocytes express three variants of AURKC that are not functionally equivalent in supporting meiosis, but fully complement meiosis when expressed simultaneously.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Actidion, from microbial, ≥94% (TLC)
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Cycloheximid -Lösung, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Anti-acetyliertes-Tubulin-Antikörper, monoklonaler Antikörper der Maus in Maus hergestellte Antikörper, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Cycloheximid, ≥95% (HPLC)
Sigma-Aldrich
Monoklonaler Anti-α-Tubulin-Antikörper in Maus hergestellte Antikörper, clone DM1A, ascites fluid
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Actidion, Biotechnology Performance Certified
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Monoklonaler Anti-grünfluoreszierendes Protein (GFP)-Antikörper in Maus hergestellte Antikörper, clone GFP-20, ascites fluid
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Milrinon, ≥97% (TLC), powder
Sigma-Aldrich
Monastrol, ≥98% (HPLC), solid
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri