Merck
  • Overexpression, purification, crystallization and preliminary X-ray crystallographic analysis of SF173 from Shigella flexneri.
Overexpression, purification, crystallization and preliminary X-ray crystallographic analysis of SF173 from Shigella flexneri.
Acta crystallographica. Section F, Structural biology communications (2015-01-24)
Ha-Neul Kim, Jeong-Gi An, Yoo-Sup Lee, Seung-Hyeon Seok, Hee-Seop Yoo, Min-Duk Seo
ZUSAMMENFASSUNG
Shigella flexneri is a Gram-negative, anaerobic bacterium in the genus Shigella that can cause diarrhoea in humans. SF173, a hypothetical protein from S. flexneri 5a strain M90T, has been cloned, overexpressed, purified and crystallized as a part of laboratory-scale structural genomics project. The SF173 protein was crystallized using the sitting-drop vapour-diffusion method in the presence of 0.8 M succinic acid pH 7.0 at 293 K. Preliminary X-ray diffraction analysis revealed that the crystal diffracted to 1.47 Å resolution and belonged to space group I432, with unit-cell parameters a=b=c=110.245 Å.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis