Merck
  • Influence of fetuin and hyaluronan on the post-thaw quality and fertilizing ability of Holstein bull semen.
Influence of fetuin and hyaluronan on the post-thaw quality and fertilizing ability of Holstein bull semen.
Cryobiology (2015-05-13)
Serpil Sarıözkan, Mustafa Numan Bucak, Pürhan Barbaros Tuncer, Serhat Büyükleblebici, Ayşe Eken, Cemal Akay
ZUSAMMENFASSUNG
It was determined that fetuin and hyaluronan supplementation did not provide any significant effect on the post-thaw subjective and CASA motility percentages and sperm motion characteristics, in comparison to the controls (P>0.05). Sperm acrosome and total abnormalities were similar in all groups (P>0.05). Groups M (hyaluronan+fetuin) and H (hyaluronan) displayed a higher rate of sperm membrane integrity, compared to that of Group C (control) (P<0.01). According to the results of the comet assay, the lowest percentage of sperm with damaged DNA was achieved in Group H, when compared to all of the experimental groups (P<0.01). Furthermore, all of the additives resulted in a lower rate of sperm with damaged DNA than that of the controls, and thus, reduced DNA damage (P<0.01). For pregnancy rates, there were no significant differences between the extender groups (P>0.05). MDA formation was found to be lower in Groups M and F (P<0.01). In Group M, SOD activity was determined to have significantly increased (23.61±5.62 U/ml) compared to the other groups (P<0.01). All experimental groups had a GSH-Px activity higher than that of the control group (P<0.01).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Supelco
DL-Dithiothreitol -Lösung, 1 M in H2O
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Formaldehyd -Lösung, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Formaldehyd-12C -Lösung, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®